Skip to Content
Merck
All Photos(1)

Key Documents

CLS354401

Corning® BioCoat Collagen I Culture Dish

diam. 60 mm

Synonym(s):

Cell Culture Dish, plastic cell culture dish, coated cell culture dish

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.13

feature

lid

packaging

pkg of 20 ea (1 pack )

manufacturer/tradename

Corning 354401

diam.

60 mm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.</p> Applications include:</p><ul><li>Promotion of cell attachment and spreading</li><li>Rapid expansion of cell populations</li><li>Serum-free or reduced serum culture</li><li>Cell adhesion assays</li><li>Improving survival of primary cells in culture</li></ul>Source</p><li>Rat tail tendon</li>Quality Control</p><ul><li>Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells</li><li>Tested and found negative for bacteria and fungi</li><li>Collagen I purity >90% by SDS-PAGE</li></ul>Storage</p><li>4°C to 30°C under dry conditions.</li>

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service