Skip to Content
MilliporeSigma

Skip To

08-115

Rat Collagen Type I

from rat tail, liquid, 2-5 mg/mL, suitable for cell culture, used for 3D gel formation

Synonym(s):

Rat tail collagen, Rat tail collagen type I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg

Estimated to ship onAugust 31, 2026

$352.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Biological source:
rat
Form:
liquid, solution
Technique(s):
cell culture | mammalian: suitable (and for 3D proliferation assays.)
Concentration:
100 mg

$352.00


Estimated to ship onAugust 31, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Collagen Type I, rat tail, 100 mg, solution, BioReagent, suitable for cell culture

biological source

rat

Quality Segment

form

liquid, solution

manufacturer/tradename

Upstate®

concentration

100 mg

technique(s)

cell culture | mammalian: suitable (and for 3D proliferation assays.)

input

sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type: mouse embryonic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rat ... COL1A1(29393)

General description

In humans, type I collagen is the most abundant and broadly expressed collagen. This heterotrimer contains two alpha 1 (I) chains and one alpha 2 (I) chain.[1] Collagen type I α 1 (COL1A1), a major component of collagen type I, codes for pro-α 1 chains of type I collagen.[2] It has repeating Gly-Xaa-Yaa tri-peptides motifs. The collagen alpha-1(I) chain protein is a member of the collagen complex superfamily.[1] COL1A1 has a von Willebrand factor type C (vWFC) domain, a collagen triple-helix repeat, and a fibrillar collagen C-terminal domain (COLF).[2]

Application

Collagen Type I, rat tail has been used:
  • for collagen 3D proliferation assays[1]
  • for atelocollagen proliferation assays[1]
  • in a comparative study to make microcapsules for templating on mesoporous cores of pure vaterite calcium carbonate (CaCO3) crystals to evaluate the structure-property relationship[2]
  • in coverslip coating to grow the human cerebral microvascular endothelial cell line (hCMEC/D3) for creating a cell coculture and chronic subdural hematoma (CSDH) patient hematoma sample induced injury model[3]

Biochem/physiol Actions

Collagen type I α 1 (COL1A1) plays a key role in chemoresistance and may act as a therapeutic target. It is involved in carboplatin-resistant ovarian cancer (OV).[1] COL1A1 serves as an oncogene in the development of colorectal cancer (CRC).[2] Dominant inherited COL1A1 gene mutations result in connective tissue disorders such as osteogenesis imperfecta (OI).[1]

Physical form

0.02N acetic acid, pH 3.67

Preparation Note

1 year at 4°C

Analysis Note

Routinely evaluated by testing for uniform gelation and for attachment of and spreading of PC-12 rat cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
11179179001234138AB765P
technique(s)

cell culture | mammalian: suitable (and for 3D proliferation assays.)

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

ELISA: suitable, immunohistochemistry: suitable, radioimmunoassay: suitable, western blot: suitable

biological source

rat

biological source

rat tail

biological source

human skin

biological source

rabbit

concentration

100 mg

concentration

-

concentration

-

concentration

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

form

liquid, solution

form

lyophilized (clear, colorless solution after reconstitution)

form

lyophilized

form

-

shipped in

wet ice

shipped in

-

shipped in

ambient

shipped in

dry ice


Still not finding the right product?

Explore all of our products under Collagen Type I, rat tail


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. Does this product require pH neutralization by addition of acid or base for gelation?

    1 answer
    1. The gelation process for this product does not require pH neutralization by the addition of acid or base. The procedure involves pipetting undiluted collagen into culture dishes, exposing them to ammonia vapor, and rinsing with sterile distilled water multiple times to remove ammonia. The final rinse is replaced with a sterile medium or balanced salt solution with serum, and the dishes are stored at 4°C until use.

      Helpful?

  2. Hi team: I hope this finds you well. May I know the concentration of collagen type I , rat tail 08-115? Much appreciated. Kind regards, Ovi

    1 answer
    1. The concentration is provided in lot specific Certificates of Analysis, typically within the range of 3 to 5 mg/ml.

      Helpful?

  3. Accidentally, I opened the collagen out of the flow chamber. Should I filter it or autoclave it to ensure the sterile conditions?

    1 answer
    1. Collagen solutions may be sterilized by dialysis in a 0.25% acetic acid and 0.5% chloroform solution.
      Avoid sterilizing the collagen solution by membrane filtration, as substantial protein loss may occur by this method. The coating of tissue culture plastic dishes may be performed by air-drying. Dried coated dishes may then be sterilized by exposure to UV light in a sterile culture hood or by rinsing with 70% ethanol.

      Helpful?

  4. Hi, what is the concentration of the collagen?

    1 answer
    1. The concentration of this product will be listed on the lot-specific Certificate of Analysis. Please see the link below to access a Certificate:
      https://www.emdmillipore.com/US/en/product/Collagen-Type-I-rat-tail,MM_NF-08-115#anchor_COA

      Helpful?

  5. Once diluted in 0.02N acetic acid, does the collagen solution require sterilisation for use in cell culture? If so, which method is the most effective: UV, autoclave or 0.22uM sterile filter (should this be PES or PVDF)?

    1 answer
    1. This product is supplied as a sterile solution in 0.02 N Acetic acid. The package size is 100 mg, however, the concentration is variable and reported on the lot-specific Certificate of Analysis. The typical range is 4 - 5 mg/mL. Dilutions can be made with sterile PBS. Further sterilization is not required. If necessary, coated slides and coverslips can be sterilized by UV.

      Helpful?

Reviews

No rating value

Active Filters