Skip to Content
Merck
All Photos(1)

Key Documents

H69

91091802, human lung, Not specified

Synonym(s):

H69 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

H69, 91091802

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Aneuploid

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung small cell carcinoma

Cell Line Description

This cell line has been established from the pleural fluid of a 55 year old Caucasian male with small cell carcinoma of the lung prior to treatment. Inoculated into athymic mice, the cells form tumours with a typical histology of the original biopsy specimen. Elevated levels of the biomarkers L-dopa decarboxylase, bombesin-like immunoreactivity, neuron-specific enolase and brain isozyme of creatine kinase were reported. Cells grow as spherical floating aggregates.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 12
D16S539: 11
D5S818: 11,13
D7S820: 9
THO1: 8,9
TPOX: 10
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 6x10,000 to 3x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service