Skip to Content
Merck
All Photos(1)

Key Documents

C9033

Sigma-Aldrich

β-Chloro-L-alanine hydrochloride

Alanine aminotransferase inhibitor

Synonym(s):

(R)-2-Amino-3-chloropropanoic acid, 3-Chloro-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6ClNO2 · HCl
CAS Number:
Molecular Weight:
160.00
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥98% (TLC)

form

powder

mp

205 °C

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Cl[H].N[C@@H](CCl)C(O)=O

InChI

1S/C3H6ClNO2.ClH/c4-1-2(5)3(6)7;/h2H,1,5H2,(H,6,7);1H/t2-;/m0./s1

InChI key

IENJPSDBNBGIEL-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Chloro-L-alanine hydrochloride has been used:
  • as a substrate for screening tyrosine phenol lyase steady state kinetics
  • in the inhibition of serine palmitoyltransferase and cytotoxicity studies in lymphocyte Ly-1 cells
  • as an inhibitor of alanine transaminase in wheat germ cell-free protein synthesis system

Biochem/physiol Actions

β-Chloro-L-alanine hydrochloride is an inhibitor of alanine aminotransferase (ALAT). Inhibition of ALAT leads to suppression of tumor progression in lung carcinoma.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxytetracycline have the therapeutic efficiency in CD133+ HCC population through suppression CD133 expression by decreasing of protein stability of CD133
Song Y, et al.
Scientific Reports, 8(1), 16100-16100 (2018)
Specificity of inhibitors of serine palmitoyltransferase (SPT), a key enzyme in sphingolipid biosynthesis, in intact cells: a novel evaluation system using an SPT-defective mammalian cell mutant
Hanada K, et al.
Biochemical Pharmacology, 59(10), 1211-1216 (2000)
A novel way of amino acid-specific assignment in 1 H-15 N HSQC spectra with a wheat germ cell-free protein synthesis system
Morita EH, et al.
Journal of Biomolecular Nmr, 30(1), 37-45 (2004)
Inhibition of alanine aminotransferase (ALAT) in silico and in vivo promotes mitochondrial metabolism to impair malignant growth
Beuster G, et al.
The Journal of Biological Chemistry, jbc-M110 (2011)
Affinity labeling of alanine aminotransferase by 3-chloro-L-alanine.
Y Morino et al.
The Journal of biological chemistry, 254(2), 279-285 (1979-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service