Skip to Content
Merck
All Photos(4)

Documents

C6288

Sigma-Aldrich

Cellulose

fibers, (medium)

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

(softwood tree pulp)

form

fibers

impurities

dark particles

color

white

particle size

(medium)

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose, a β-glucan insoluble in water is an important constituent of plant cell wall. It is a tough, fibrous polysaccharide, which has a linear chain of glucose residues, linked with β-1,4 bond. Plants produces this biopolymer abundantly.

Application

Cellulose (medium fibers) is used in metabolic pathway and carbohydrate applications. Cellulose has been used to study biofuel and biorefinery applications.
Cellulose has been used:
  • for biomass analysis in pyrolysis process
  • in column chromatography to purify organisms from uninfected erythrocytes and white cells
  • as a standard in fourier transform infrared spectroscopy (FTIR) to study conformational characteristics of bacterial cellulose (BC) obtained from Hestrin-Schramm (HS) medium
  • as a control in diet-related animal studies to evaluate the hypocholesterolemic activity of botryosphaeran
High purity cellulose powders for partition chromatography.

Biochem/physiol Actions

Cellulose helps in maintaining the structural stability of plant cell walls. It is an important component of paper and fabrics made from cotton, and linen.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trichoderma Enzymes for Textile Industries
Biotechnology and Biology of Trichoderma, 351-362 (2014)
Comparative studies on the pyrolysis of cellulose, hemicellulose, and lignin based on combined kinetics
Yeo JY, et al.
Journal of the Energy Institute (2017)
Visualization of Cellobiohydrolase I from Trichoderma reesei Moving on Crystalline Cellulose Using High-Speed Atomic Force Microscopy
Test, 510, 169-182 (2012)
Abhinay Ramaprasad et al.
BMC biology, 19(1), 69-69 (2021-04-24)
Rodent malaria parasites (RMPs) serve as tractable tools to study malaria parasite biology and host-parasite-vector interactions. Among the four RMPs originally collected from wild thicket rats in sub-Saharan Central Africa and adapted to laboratory mice, Plasmodium vinckei is the most
Biopolymers: Biodegradable Alternatives to Traditional Plastics
Green Chemistry, 753-770 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service