biological source
plant
Quality Level
form
powder
color
white
storage temp.
room temp
SMILES string
O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO
InChI
1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1
InChI key
GUBGYTABKSRVRQ-WFVLMXAXSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Cellulose, a polysaccharide composed of long chains of β(1,4) linked D-glucose units, is used in a wide range of applications that require a carbohydrate structural back-bone such microfibrils, biofilms, and biodegradable composites. It may be used for the preparation of chromatographic structural media, in electrophoresis gels and ionic separations and as a substrate to identify and study cellulosic enzymes such as endoglucanases and β-glucosidase(s) involved with cellulosic systems. Cellulose may be used in saccharification and ethanol fermentation research.
Other Notes
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bartosz Fotschki et al.
Nutrients, 12(9) (2020-09-17)
In recent years, dietary products with quinoa and buckwheat have attracted attention mostly due to the high nutritive value of their protein fraction. However, their dietary effect on intestinal microbiota activity and related systemic responses are still poorly understood. Therefore
Arezki Boudaoud et al.
Nature protocols, 9(2), 457-463 (2014-02-01)
Cell biology heavily relies on the behavior of fibrillar structures, such as the cytoskeleton, yet the analysis of their behavior in tissues often remains qualitative. Image analysis tools have been developed to quantify this behavior, but they often involve an
Rossella Guerrieri et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(34), 16909-16914 (2019-08-07)
Multiple lines of evidence suggest that plant water-use efficiency (WUE)-the ratio of carbon assimilation to water loss-has increased in recent decades. Although rising atmospheric CO2 has been proposed as the principal cause, the underlying physiological mechanisms are still being debated
Ekaterina M Podgorbunskikh et al.
Polymers, 11(7) (2019-07-22)
As a heterogeneous process, enzymatic hydrolysis depends on the contact area between enzymes and the cellulose substrate. The surface area of a substrate is typically evaluated through the sorption of gases (nitrogen, argon, or water vapor) or sorption of high-molecular-weight
Neha Aggarwal et al.
Journal of biomedical materials research. Part A, 102(12), 4224-4233 (2014-01-28)
Here, we report fabrication of polyelectrolyte multilayers by blending a natural glycosaminoglycan (heparin) with semisynthetic cellulose sulfates as polyanions paired with polycation chitosan. Two types of polyanionic blends were prepared by mixing heparin with either cellulose sulfates (CS) of high
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C8002-1KG | 04061835366545 |
| C8002-5KG | 04061833521434 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service