Skip to Content
Merck
All Photos(1)

Key Documents

C4648

Sigma-Aldrich

Corn steep liquor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

corn

form

suspension

technique(s)

microbiological culture: suitable

storage temp.

2-8°C

General description

Corn steep liquor, a by-product of the corn wet-milling industry consists of 50% water and several ingredients like, sugars and proteins. It is a viscous liquid by-product with a pH of 3.7 to 4.1.

Application

Corn steep liquor has been used:
  • as a component in broth to act as indicator for starch utilizationas a component in fermentation medium, low-cost medium and in LB medium for the simultaneous saccharification and co-fermentation (SSCF) test
  • as a component in fermentation medium for optimization studies

Corn steep liquor is useful as an inexpensive alternative to peptone for a wide variety of microbiological production methods, including fed-batch production of recombinant proteins in E. coli., high density culture of S. cerevisiae, and fermentative production of lactic acid.

Biochem/physiol Actions

Corn steep liquor can be used as a best alternative for yeast extract, as a nitrogen source in biochemical industries. The vitamins, minerals and nitrogen present in the liquor can be used for fermentation.

Other Notes

A viscous concentrate of corn solubles, rich in vitamins, amino acids, minerals and other growth stimulants. Contains approx. 50% (w/w) solids.
A viscous concentrate of corn solubles, rich in vitamins, amino acids, minerals and other growth stimulants. Contains approx. 50% (w/w).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyeong-U Son et al.
Experimental and therapeutic medicine, 6(4), 909-912 (2013-10-19)
The aim of this study was to investigate whether S-(-)-10,11-dihydroxyfarnesic acid methyl ester produced by cell subtype
The effect of temperature and Lactobacillus amylovorus and Lact. plantarum, applied at ensiling, on wheat silage
Weinberg ZG, et al.
Journal of Applied Microbiology, 84(3), 404-408 (1998)
Livia Martinelli et al.
Extremophiles : life under extreme conditions, 21(4), 755-773 (2017-05-14)
Halophilic fungal strains isolated from historical wooden staircase in a salt mine in Austria, and from wall biofilm and soil of a cave in the Coastal Range of the hyperarid Atacama Desert in Chile were characterised and described newly as
Optimization of the associative growth of novel yoghurt cultures in the production of biomass, beta-galactosidase and lactic acid using response surface methodology
Tari C, et al.
International dairy journal, 19(4), 236-243 (2009)
EVAPORATION vert Uses in the Food Industry
Encyclopedia of Food Sciences and Nutrition (Second Edition), 2205-2211 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service