Skip to Content
Merck

C6304

Corn meal

BioReagent, suitable for insect cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352205
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

maize

Quality Segment

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable

shipped in

ambient

storage temp.

room temp

Other Notes

May also be used in corn-meal agar for studies of yeast differentiation.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library