Skip to Content
Merck

C8740

CelLytic B Cell Lysis Reagent

For bacterial cell lysis, 10× concentrate

Synonym(s):

bacterial cell lysis reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116134
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

Application

A proprietary, non-denaturing formulation of zwitterionic detergents used for the lysis of bacterial cells and extraction of recombinant proteins.
TM[="CelLytic"] B Cell Lysis Reagent has been used for lysing E.coli 5-helix bundle protein. It is also suitable for lysing hepatitis B X-interacting protein (HBXIP) and E. coli TG1 cells.

Features and Benefits

  • Higher protein extraction efficiency than traditional methods such as sonication and lysozyme
  • Scalable for 1 to 25 grams of bacterial cell paste
  • No interference with downstream applications such as affinity chromatography, IP, and Western blotting
  • Compatible with protease inhibitors, inhibitor cocktails, chaotropes, salts, chelating agents and reducing agents

Other Notes

10X concentrate of detergent blend contains no buffering components, giving the researcher flexibility to integrate the lysis reagent into their desired system. Also amenable to automation protocols for recombinant protein extraction and purification.

Legal Information

Covered by US Patent No 7,282,475 B2 and are sold for research use only. Commercial use requires addtional licenses.
CelLytic is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under CelLytic B Cell Lysis Reagent


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library