Skip to Content
Merck
All Photos(1)

Documents

C2999

Sigma-Aldrich

CryoStor® cell cryopreservation media

CS5

Synonym(s):

cell freezing medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable
cryopreservation: suitable

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The CryoStor CS2, CS5, and CS10 family of preservation solutions represents the next generation of cryopreservation media. Designed to prepare and preserve cells in ultra low temperature environments (-80 to -196 °C), CryoStor media provide a safe, protective environment for cells and tissues during the freezing, storage, and thawing process. Through modulating the cellular biochemical response to the cryopreservation process, these media provide enhanced cell viability and functionality, while eliminating the need to include serum, proteins, or high levels of cytotoxic agents.

CryoStor CS2, CS5, and CS10 are a series of cell specific, optimized preservation media, uniquely formulated to address the molecular biological aspects of cells during the cryopreservation process; thereby, directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.

These media are recommended for the preservation of stem cells, hepatocytes, tissue samples, and other extremely sensitive cell types.

Application

CryoStor, a series of cell-specific, optimized preservation media, is uniquely formulated to address the molecularbiological aspects of cells during the cryopreservation process thereby directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.
Cryostor CS5 is formulated to contain 5% dimethyl sulfoxide (DMSO). Recommended for cryopreservation of most cell types.

Cryostor® Cryopreservation Video Protocol

Other Notes

Formulation contains 5% DMSO.

Legal Information

CryoStor is a registered trademark of BioLife Solutions, Inc.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Li et al.
Journal of virology (2021-03-05)
Previously, we showed that substitution of HIV-1 Env residue 375-Ser by bulky aromatic residues enhances binding to rhesus CD4 and enables primary HIV-1 Envs to support efficient replication as simian-human immunodeficiency virus (SHIV) chimeras in rhesus macaques (RMs). Here, we
Ryan S Roark et al.
Science (New York, N.Y.), 371(6525) (2020-11-21)
Neutralizing antibodies elicited by HIV-1 coevolve with viral envelope proteins (Env) in distinctive patterns, in some cases acquiring substantial breadth. We report that primary HIV-1 envelope proteins-when expressed by simian-human immunodeficiency viruses in rhesus macaques-elicited patterns of Env-antibody coevolution very
Dominic M Clarke et al.
Cytotherapy, 11(4), 472-479 (2009-06-06)
Peripheral blood stem cells (PBSC) have become the preferred stem cell source for autologous hematopoietic transplantation. A critical aspect of this treatment modality is cryopreservation of the stem cell products, which permits temporal separation of the PBSC mobilization/collection phase from
J Stylianou et al.
Cytotherapy, 8(1), 57-61 (2006-04-22)
Hematopoietic stem cells (HSC) have traditionally been frozen using the cryoprotectant DMSO in dextran-40, saline or albumin. However, the process of freezing and thawing results in loss of HSC numbers and/or function. This study investigated the use of CryoStor for
Haichen Niu et al.
Brain pathology (Zurich, Switzerland), 30(6), 1102-1118 (2020-07-18)
Olfactory dysfunction is one of the early symptoms seen in Parkinson's disease (PD). However, the mechanisms underlying olfactory pathology that impacts PD disease progression and post-mortem appearance of alpha-Synuclein (α-Syn) inclusions in and beyond olfactory bulb in PD remain unclear.

Articles

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Protocols

Cryopreservation affects post-thaw recovery, viability, and functionality. Stress during freezing and suboptimal media lead to cell death.

Cryopreservation affects post-thaw recovery, viability, and functionality. Stress during freezing and suboptimal media lead to cell death.

Cryopreservation affects post-thaw recovery, viability, and functionality. Stress during freezing and suboptimal media lead to cell death.

Cryopreservation affects post-thaw recovery, viability, and functionality. Stress during freezing and suboptimal media lead to cell death.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service