Skip to Content
Merck

Skip To

D2438

Dimethyl sulfoxide

sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma

Synonym(s):

DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
UNSPSC Code:
12191502
NACRES:
NA.75
PubChem Substance ID:
EC Number:
200-664-3
Beilstein/REAXYS Number:
506008
MDL number:
Grade:
BioPerformance Certified
Technique(s):
cell culture | hybridoma: suitable, cryopreservation: suitable
Bp:
189 °C (lit.)
Vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

BioPerformance Certified

Quality Segment

agency

USP XXIV, meets EP testing specifications, meets USP testing specifications

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

sterility

sterile-filtered

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

storage condition

(well-ventilated place. Keep cool.)

technique(s)

cell culture | hybridoma: suitable, cryopreservation: suitable

impurities

≤0.1% water

color

colorless

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

solubility

H2O: miscible (completely)

density

1.10 g/mL (lit.)

storage temp.

room temp

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

General description

Dimethyl sulfoxide (DMSO) is an aprotic solvent. Therapeutic and toxic molecules that are not soluble in water are often soluble in DMSO. Liquid nitrogen used in conjunction with a cryoprotective agent such as DMSO is a widely used method for preserving cells. DMSO is used to prepare stock solutions of various molecules and is also used as a control for these molecules. It has been used in cell migration assay for dissolving post-migration cells. BPC-tested DMSO also meets the requirements of biotechnology and tissue engineering applications.

Application

Dimethyl sulfoxide has been used as a vehicle control to culture and induce hepatic stellate cells (HSCs) activation in vivo. It has also been used to prepare a stock solution to study the cytotoxicity of ferulic acid (FA) on nucleus pulposus (NP) cells.

Features and Benefits

Ready-to-use sterile filtered product conveniently packaged in septum screw-capped amber bottles. Meets USP and EP testing requirements

Disclaimer

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
41639PHR130941640
description

sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma

description

BioUltra, Molecular Biology, ≥99.5% (GC)

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

puriss. p.a., ACS reagent, ≥99.9% (GC)

grade

BioPerformance Certified

grade

Molecular Biology

grade

pharmaceutical secondary standard, certified reference material

grade

ACS reagent, puriss. p.a.

technique(s)

cryopreservation: suitable, cell culture | hybridoma: suitable

technique(s)

GC/GC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

GC/GC: suitable

Quality Level

400

Quality Level

200

Quality Level

300

Quality Level

300

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

solubility

-

solubility

-

form

liquid

form

liquid

form

-

form

liquid

refractive index

n20/D 1.479 (lit.)

refractive index

n20/D 1.479

refractive index

-

refractive index

n20/D 1.479


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide


Storage Class

10 - Combustible liquids

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 17 Questions  
  1. Is it okay to mix it with freezing media that is kept refrigerated? Additionally, when I mixed DMSO with freezing media that was kept refrigerated, a white floating substance appeared. Do you know what might have caused this?

    1 answer
    1. The DMSO is stable at room or refrigerated temperatures. Typically, cell freezing media containing serum and DMSO, are stable for at least several days when thawed and refrigerated. Other constituents of freezing medium, primarily serum, may contribute to the appearance of particulates. For detailed troubleshooting, please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information so that a member of the Technical Service team can reach out to assist further.

      Helpful?

  2. For D2438-5X10ML, do they come in glass ampoule or bottle with lid?

    1 answer
    1. Yes, both package sizes have a septum cap closure as shown in the image below:

      Helpful?

  3. Hi, what is the difference between DMSO product# D2438 and D2650? What is Hybri-Max vs. BioPerformance certified and meets USP testing specification?

    1 answer
    1. Both D2438 and D2650 are suitable for cell culture. The D2650 is part of the Hybri-Max™ product line. Hybri-Max products have been tested for use in hybridoma creation and culture. The D2438 is part of the BioPerformance Certified group of products. These products are tested and optimized for use in biological applications. For this product and a number of others, this includes testing to meet USP and EP guidelines. Please see the links below to review a sample Certificate of Analysis for each item.
      D2650
      https://www.sigmaaldrich.com/product/sigma/d2438#product-documentation
      D2438
      https://www.sigmaaldrich.com/product/sigma/d2650#product-documentation

      Helpful?

  4. Purity grade of the product ?

    1 answer
    1. The purity of this product is listed on the lot specific Certificate of Analysis. Please navigate to the DOCUMENTATION section of the Product Detail Page to access a Certificate:
      https://www.sigmaaldrich.com/product/sigma/d2438#product-documentation

      Helpful?

  5. El DMSO es sensible a la luz, hay que tomar alguna precaución al respecto?

    1 answer
    1. This product is packaged in amber glass and is protected from light. The 10 mL size is packaged in a sealed ampule. Once opened, any content that is not used should also be stored in amber glass. For long term storage, the material may also be stored under inert gas to preserve the shelf life. Note that this material has a tendency to absorb water. Containers should be tightly sealed.

      Helpful?

  6. what are the storing conditions?

    1 answer
    1. This product is stored at room temperature. Store in a well-ventilated place. Keep cool. Please see the Material Safety Data Sheet for more information: https://www.sigmaaldrich.com/sds/sigma/d2438.

      Helpful?

  7. What is the difference between products D2438 and D8418?

    1 answer
    1. The D2438 is a BioPerformance Certified grade product suitable for cell culture, hybridoma creation, and cryopreservation. It is intended for research use only but does meet USP and EP testing requirements. The D8418 is a molecular biology-grade product suitable for DNA sequencing, PCR, and transfection. It has been tested for RNAse and DNAse. The products can be selected based on the application.

      Please view the sample Certificate of Analysis for each product to see the testing specifications associated with that item:
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/PDF734068.pdf
      https://www.sigmaaldrich.com/coa/SIGMA/D2438/RNBL8837

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  9. What is the DMSO dissolved in?

    1 answer
    1. DMSO is not dissolved in any solvent. It is a pure chemical compound that is a liquid at room temperature.

      Helpful?

  10. What is the basis for the endotoxin specification for Dimethyl sulfoxide (DMSO)?

    1 answer
    1. The industry standard for 'endotoxin free' is less than 1 EU/mL.

      Helpful?

1–10 of 17 Questions  

Reviews

Active Filters

  1. 1 Ratings-only review