Skip to Content
Merck
All Photos(2)

Key Documents

C2648

Sigma-Aldrich

Cholesterol 5β,6β-epoxide

≥98%

Synonym(s):

5β,6β-Epoxycholestan-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.28

Assay

≥98%

form

powder

functional group

epoxy

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)[C@]3(O4)[C@H]4C2)([H])CC[C@@]5(C)[C@@]1([H])CC[C@]5([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24-27(29-24)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28H,6-16H2,1-5H3

InChI key

PRYIJAGAEJZDBO-UHFFFAOYSA-N

Application

Cholesterol 5β,6β-epoxide was used to treat PC12 cells or HepG2 and Caco-2 cells to study the effects of cholesterol oxidation.

Biochem/physiol Actions

Cholesterol 5β,6β-epoxide is an oxysterol produced as an oxidation product of cholesterol. Oxysterols are cytotoxic and induce death in monocytes, smooth muscle cells and endothelial cells. The mechanism of apoptosis induced by oxysterols may involve caspases or DNA fragmentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Hua Chen et al.
FEBS letters, 580(2), 479-483 (2006-01-03)
The adaptive response induced by the lipid peroxidation products, such as phosphatidylcholine hydroperoxide, lysophosphatidylcholine (LysoPC), 15-deoxy-Delta(12,14)-prostaglandin J(2), 4-hydroxynonenal (4-HNE), hydroxyoctadecadienoic acid, 7-hydroxycholesterol, and cholesterol 5beta,6beta-epoxide, was investigated in this study. Although these products have been implicated in oxidative stress-related diseases
Cholesterol oxides as biomarkers of oxidative stress in type 1 and type 2 diabetes mellitus.
Ferderbar S, Pereira EC, Apolinario E, et al.
Diabetes and Metabolism, 23, 35-42 (2007)
A J O'Sullivan et al.
Journal of applied toxicology : JAT, 23(3), 191-197 (2003-06-10)
Cholesterol can be oxidized to form a variety of cholesterol oxidation products also known as oxysterols. The aims of the present study were to compare the cytotoxic effects of four oxysterols, namely 25-hydroxycholesterol (25-OHC), 7beta-hydroxycholesterol (7beta-OHC), cholesterol-5beta,6beta-epoxide (beta-epox) and cholesterol-5alpha,6alpha-epoxide
S Ferderbar et al.
Diabetes/metabolism research and reviews, 23(1), 35-42 (2006-04-25)
Oxidative stress plays an important role in the pathophysiology of diabetes mellitus. The aim of this study was to evaluate the formation of cholesterol oxides (ChOx) as biomarkers of oxidative stress in subjects with impaired glucose tolerance (IGT) and diabetes.
D U Ahn et al.
Poultry science, 78(7), 1060-1064 (1999-07-15)
A study was conducted to develop a solvent system that will clean egg yolk samples and concentrate cholesterol oxides effectively before analysis. Cholesterol oxide standards or lipid samples (0.2 g) loaded onto a silicic column were washed with a portion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service