Skip to Content
Merck
All Photos(4)

Key Documents

C2394

Sigma-Aldrich

5-Cholesten-3β-ol-7-one

≥90%

Synonym(s):

3β-Hydroxy-5-cholesten-7-one, 7-Ketocholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
MDL number:
UNSPSC Code:
41141804
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥90%

form

powder

functional group

ketone

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@@]([C@](CC[C@H](O)C3)(C)C3=CC4=O)([H])[C@]4([H])[C@]1([H])CC[C@]2([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-23,25,28H,6-15H2,1-5H3/t18-,20+,21-,22+,23+,25+,26+,27-/m1/s1

InChI key

YIKKMWSQVKJCOP-ABXCMAEBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Cholesten-3β-ol-7-one was used as a standard in the determination of cholesterol derivatives by GC-MS and HPLC.

Biochem/physiol Actions

5-Cholesten-3β-ol-7-one is a cholesterol derivative that decreases the membrane potential of lipid bilayers. It reportedly accumulates in atherosclerotic plaques that leads to apoptosis of vascular cells.

Preparation Note

5-Cholesten-3β-ol-7-one yields clear to slightly hazy, colorless to faint yellow solution in chloroform at 250 mg/5 ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Starke-Peterkovic et al.
Biophysical journal, 90(11), 4060-4070 (2006-03-04)
The effect of cholesterol removal by methyl-beta-cyclodextrin on the dipole potential, psi(d), of membrane vesicles composed of natural membrane lipids extracted from the kidney and brain of eight vertebrate species was investigated using the voltage-sensitive fluorescent probe di-8-ANEPPS. Cyclodextrin treatment
C Adcox et al.
Journal of agricultural and food chemistry, 49(4), 2090-2095 (2001-04-20)
The cytotoxicity of cholesterol and a mixture of beta-sitosterol/campesterol (50%/40%) and their oxides was examined in a cultured-derived macrophage cell line, C57BL/6. Cell numbers, lactate dehydrogenase (LDH) leakage, protein content, lipid uptake, and mitochondria dehydrogenase activity were determined after exposure
E Boselli et al.
Journal of chromatography. A, 917(1-2), 239-244 (2001-06-14)
Pressurized liquid extraction (PLE, ASE) was compared with the Folch procedure (a solid-liquid extraction with chloroform/methanol 2:1, v/v) for the lipid extraction of egg-containing food; the accuracy of PLE for the quantitative determination of oxysterols in whole egg powder was
Aline Yammine et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-18)
The brain, which is a cholesterol-rich organ, can be subject to oxidative stress in a variety of pathophysiological conditions, age-related diseases and some rare pathologies. This can lead to the formation of 7-ketocholesterol (7KC), a toxic derivative of cholesterol mainly
Anne Pilotelle-Bunner et al.
Biochimica et biophysica acta, 1787(8), 1039-1049 (2009-03-25)
Three different cholesterol derivatives and phloretin, known to affect the local electric field in phospholipid membranes, have been introduced into Rhodobacter sphaeroides reaction centre-containing phospholipid liposomes. We show that cholesterol and 6-ketocholestanol significantly slow down the interquinone first electron transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service