Skip to Content
Merck
All Photos(2)

Key Documents

C25120

Sigma-Aldrich

CM Sephadex® C-25

Synonym(s):

CM Sephadex®, Carboxymethyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

powder

Quality Level

technique(s)

activity assay: suitable

matrix

cross-linked dextran

bead size

40-125 μm (dry)

pore size

~30,000 Da exclusion limit

operating pH

6-10

pH

2—12

capacity

4.0-5.0 meq/g ion exchange capacity

compatibility

mode of use weak cation exchange chromatography

General description

C25120-100G′s updated product number is GE17-0210-01

Application

Sephadex® C-25 is used in ion exchange chromatography, cation exchange media, protein chromatography, separation media and resins. Sephadex® C-25 has been used to study wasp envenomation and to demonstrate that snake protease inhibitors and neurotoxic homologs originated from a common ancestor.

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mononuclear and dinuclear ruthenium(III) and ruthenium(IV) complexes of 1,4-dicyanamido-2,5-dimethylbenzene dianion.
Naklicki, M.L., et al.
Inorgorganica Chimica Acta, 242(1-2), 63-69 (1996)
Junyou Han et al.
Toxicon : official journal of the International Society on Toxinology, 51(5), 914-922 (2008-03-01)
Wasp is an important venomous animal that can induce human fatalities. Coagulopathy is a clinical symptom after massive wasp stings, but the reason leading to the envenomation manifestation is still not known. In this paper, a toxin protein is purified
Metal-Ligand Coupling Elements and Antiferromagnetic Superexchange in Ruthenium Dimers.
Naklicki, M.L., et al.
Inorganic Chemistry, 37(8), 1880-1885 (1998)
J W Adelson et al.
The Journal of biological chemistry, 261(23), 10569-10575 (1986-08-15)
Chymodenin, a hormone-like duodenal peptide which rapidly alters the proportions of secreted pancreatic digestive enzymes to a mixture relatively richer in chymotrypsinogen than that found in basal secretion, has been purified to homogeneity. The starting material was an acidic methanol-soluble
Cleonice C Santos et al.
Toxicology in vitro : an international journal published in association with BIBRA, 42, 54-60 (2017-04-11)
Aminochrome has been suggested as a more physiological preclinical model capable of inducing five of the six mechanisms of Parkinson's Disease (PD). Until now, there is no evidence that aminochrome induces glial activation related to neuroinflammation, an important mechanism involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service