Skip to Content
Merck
All Photos(3)

Key Documents

G100120

Sigma-Aldrich

Sephadex® G-100

Medium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

Quality Level

form

slurry

technique(s)

LPLC: suitable

matrix

Agarose

matrix active group

polymer

swelling

1 g swells to 15-20 mL gel

bead size

40-120 μm

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® is composed of repeating glucose units attached by α-1,6 glucosidic bonds. This structure ensures more aptamer binding sites and greater binding capacity.[1] It is suitable for large-scale purification and is considered stable and can be utilized more than once.
Fractionation Range (MW)
Globular Proteins: 4,000 - 150,000
Dextrans: 1,000 - 100,000

Application

Sephadex®G-100 is a gel filtration medium used in protein chromatography, affinity chromatography and gel filtration chromatography
It has been used:
  • in gel filtration column for α-amylase purification[2]
  • in gel filtration column for the purification of polysaccharidic fraction extracted from the skin of the ray Raja radula [3]
  • to remove excess protamine by size exclusion from cross-linked iron oxide (CLIO)-protamine-linked (B) peptides[4]

Other Notes

G100120-100G′s updated product number is GE17-0060-01

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed K A El-Sayed et al.
International journal of biological macromolecules, 132, 1274-1281 (2019-04-07)
AmyLa α-amylase gene from Laceyella sp. DS3 was heterologously expressed in E. coli BL21. E. coli BL21 maximally expressed AmyLa after 4 h of adding 0.02 mM IPTG at 37 °C. The recombinant AmyLa α-amylase was purified 2.19-fold through gel filtration and ion
Labeling of immune cells for in vivo imaging using magnetofluorescent nanoparticles
Pittet MJ, et al.
Nature Protocols, 1(1), 73-73 (2006)
Mohamed Ben Mansour et al.
Thrombosis research, 123(4), 671-678 (2008-07-12)
The polysaccharide fraction from the skin of the ray Raja radula was extracted, characterized and assayed for anticoagulant activity. A whole polysaccharidic fraction was extracted from the skin of the ray Raja radula by papain digestion followed by cetylpyridinium chloride
Molecular Basis of the Difference in Oxygen Affinity Between Maternal and Foetal Red Blood Cells in the Viviparous Garter Snake <I>Thanmophis Elegans</I>
Berner, N.J., and Ingermann, R.L.
The Journal of Experimental Biology, 140, 147-153 (1988)
Gene sequence, modeling, and enzymatic characterization of alpha-amylase AmyLa from the thermophile Laceyella sp. DS3
El-Sayed AKA, et al.
Starch, 69(5-6), 1600255-1600255 (2017)

Questions

1–3 of 3 Questions  
  1. My Sephadex® G-100 column is cracking. What is the maximum operating pressure that Product G100120 can take?

    1 answer
    1. The maximum operating pressure for this gel is 96 cm water = 1.6 psi.

      Helpful?

  2. How do I determine which size to use for Product G100120, Sephadex® G-100 columns?

    1 answer
    1. Generally, larger particles offer faster flow rates than finer particles, but poorer resolution.

      Helpful?

  3. What is the maximum flow rate for Product G100120, Sephadex® G-100 columns?

    1 answer
    1. The maximum flow rate is 4.2 ml/min. Data are based on columns of 2.5 cm diameter with a bed height of 30 cm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service