Skip to Content
Merck
All Photos(1)

Key Documents

S4027

Sigma-Aldrich

Sulfoxyethyl cellulose Fast Flow

fibers

Synonym(s):

Sulfoxyethyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

fibers

Quality Level

Looking for similar products? Visit Product Comparison Guide

Application

Sulfoxyethyl cellulose is used in protein chromatography, ion exchange chromatography and cation exchange media. Sulfoxyethyl cellulose has been used to purify and characterize a phosphatidylinositol transfer protein from human platelets and to purify an acid DNase (DNase II) from porcine spleen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Y George et al.
Biochimica et biophysica acta, 836(2), 176-184 (1985-09-11)
We report the purification of a phospholipid transfer protein from human platelets. This protein preferentially transfers phosphatidylinositol, with phosphatidylcholine and phosphatidylglycerol being transferred to a lesser extent. Phosphatidylethanolamine is not transferred. Transfer activity is detected by measuring the transfer of
J Hall et al.
Biochemistry, 28(6), 2568-2571 (1989-03-21)
The reaction of Rhodobacter sphaeroides cytochrome c2 with the Rb. sphaeroides cytochrome bc1 complex was studied by using singly labeled cytochrome c2 derivatives. Cytochrome c2 was treated with chlorodinitrobenzoic acid to modify lysine amino groups to negatively charged carboxydinitrophenyllysines and
Chemical structure of flavonoid compounds in wheat (<I>Triticum aestivum</I> L.) flour that contribute to the yellow colour of Asian alkaline noodles.
Asenstorfer, R.E., et al.
Journal of Cereal Science, 43(1), 108-119 (2006)
D Wong et al.
Microbial ecology, 47(1), 80-86 (2004-07-21)
Previous studies have shown that sulfate-reduction activity occurs in a heterogeneous manner throughout the terrestrial subsurface. Low-activity regions are often observed in the presence of clay minerals. Here we report that clays inhibit sulfate reduction activity in sediments and in
X Zeng et al.
Biotechnology progress, 15(6), 1003-1019 (1999-12-10)
As a result of the convective flow of solutes through porous membranes, membrane chromatography has a higher capture efficiency and a higher productivity than column chromatography and shows most promising industrial applications for the recovery, isolation, and purification of proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service