Skip to Content
Merck
All Photos(3)

Key Documents

B8686

Sigma-Aldrich

(+)-N-3-Benzylnirvanol

≥98% (HPLC), powder, CYP2C19 inhibitor

Synonym(s):

(5S)-5-Ethyl-5-phenyl-3-(phenylmethyl)-2,4-imidazolidinedione, 5-Ethyl-5-phenyl-3-(phenylmethyl)hydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O2
CAS Number:
Molecular Weight:
294.35
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Product Name

(+)-N-3-Benzylnirvanol, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CC[C@]1(NC(=O)N(Cc2ccccc2)C1=O)c3ccccc3

InChI

1S/C18H18N2O2/c1-2-18(15-11-7-4-8-12-15)16(21)20(17(22)19-18)13-14-9-5-3-6-10-14/h3-12H,2,13H2,1H3,(H,19,22)/t18-/m0/s1

InChI key

ZMZDHUHMXXALFX-SFHVURJKSA-N

Application

(+)-N-3-Benzylnirvanol may be used as a cytochrome P450 2C19 (CYP2C19) inhibitor in a comparison study to treat cocktail and individual substrates for the screening for potent cytochrome P450 (CYP) inhibition. It may also be used in comparative studies to investigate its potential as CYP and non-CYP selective inhibitors of CYP2C19 in suspended human hepatocytes.

Biochem/physiol Actions

(+)-N-3-Benzyl-nirvanol is a potent selective CYP2C19 inhibitor.
(+)-N-3-Benzyl-nirvanol is a potent selective CYP2C19 inhibitor. CYP2C19 has a high frequency of drug resistance; highly polymorphic.
(+)-N-3-Benzylnirvanol serves as an inhibitor of cytochrome P450 2B6 (CYP2B6).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James P Driscoll et al.
Chemical research in toxicology, 20(10), 1488-1497 (2007-09-26)
The hypothesis that the psychological side effects associated with the anesthetic phencyclidine (PCP) may be caused by irreversible binding of PCP or its reactive metabolite(s) to critical macromolecules in the brain has resulted in numerous in vitro studies aimed at
Guru R Valicherla et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(12), 1396-1402 (2019-02-13)
1. A protocol has been developed and validated for the high-throughput screening of eight major human cytochrome P450 (CYP) isozymes inhibition (CYP 1A2, 2C9, 2C19, 2D6, 3A4, 2B6, 2C8 and 2E1) using an in vitro probe cocktail containing eight substrates
Dustyn A Barnette et al.
Biochemical pharmacology, 170, 113661-113661 (2019-10-13)
Terbinafine N-dealkylation pathways result in formation of 6,6-dimethyl-2-hepten-4-ynal (TBF-A), a reactive allylic aldehyde, that may initiate idiosyncratic drug-induced liver toxicity. Previously, we reported on the importance of CYP2C19 and 3A4 as major contributors to TBF-A formation. In this study, we
Marie-Lynn Cuypers et al.
Drug metabolism and disposition: the biological fate of chemicals, 48(11), 1121-1128 (2020-08-26)
Early assessment of metabolism pathways of new chemical entities guides the understanding of drug-drug interactions. Selective enzyme inhibitors are indispensable in CYP reaction phenotyping. The most commonly applied CYP2C19 inhibitor, omeprazole, lacks selectivity. Two promising alternatives, (+)-N-3-benzylnirvanol and (-)-N-3-benzylphenobarbital, are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service