Skip to Content
Merck
All Photos(1)

Documents

B6688

Sigma-Aldrich

BMS 493

≥98% (HPLC), powder, pan-RAR inverse agonist

Synonym(s):

(E)-4-[2-[5,6-Dihydro-5,5-dimethyl-8-(2-phenylethynyl)naphthalen-2-yl]ethen-1-yl]benzoic acid, 4-[(1E)-2-[5,6-Dihydro-5,5-dimethyl-8-(phenylethynyl)-2-naphthalenyl]ethenyl]-benzoic acid, BMS204, 493

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24O2
CAS Number:
Molecular Weight:
404.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

BMS 493, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow

solubility

DMSO: ≥20 mg/mL

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

CC1(C)CC=C(C#Cc2ccccc2)c3cc(\C=C\c4ccc(cc4)C(O)=O)ccc13

InChI

1S/C29H24O2/c1-29(2)19-18-24(14-10-21-6-4-3-5-7-21)26-20-23(13-17-27(26)29)9-8-22-11-15-25(16-12-22)28(30)31/h3-9,11-13,15-18,20H,19H2,1-2H3,(H,30,31)/b9-8+

InChI key

YCADIXLLWMXYKW-CMDGGOBGSA-N

General description

BMS 493 inhibits the formation of neuritic processes and plasmenyethanolamine -phospholipase A2 (PLA2) activity.

Application

BMS 493 has been used:
  • as an inhibitor for the dietary and pharmacologic manipulation of retinoic acid (RA) activity in vivo and in vitro
  • for human induced pluripotent stem cells (iPSCs) culture and ventricular cardiomyocytes (VCMs) differentiation
  • to inhibit retinoic acid (RA) signaling in explants
  • as a retinoic acid receptor (RAR) inhibitor for the induction of synaptonemal complex protein 3 (SCP3) and ATP-dependent RNA helicase (DDX4) in primordial germ cells (PGCs)

Biochem/physiol Actions

BMS 493 is an inverse pan-RAR agonist. Retinoic acid receptors (RARs) are ligand-dependent transcription factors that control a number of physiological processes. RARs exert their functions by regulating gene networks controlling cell growth, differentiation, survival, and death.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Excitable dynamics and Yap-dependent mechanical cues drive the segmentation clock
Hubaud A, et al.
Cell, 171(3), 668-682 (2017)
Retinoic acid signaling is essential for airway smooth muscle homeostasis
Chen F, et al.
JCI insight, 3(16) (2018)
Spatial Fold Change of FGF Signaling Encodes Positional Information for Segmental Determination in Zebrafish
Simsek MF and Ozbudak EM
Cell Reports, 24(1), 66-78 (2018)
Jiaying Zeng et al.
Acta neuropathologica, 133(6), 967-982 (2017-01-29)
Deposition of amyloid β protein (Aβ) to form neuritic plaques in the brain is the unique pathological hallmark of Alzheimer's disease (AD). Aβ is derived from amyloid β precursor protein (APP) by β- and γ-secretase cleavages and turned over by
Structural and electrophysiological dysfunctions due to increased endoplasmic reticulum stress in a long-term pacing model using human induced pluripotent stem cell-derived ventricular cardiomyocytes
Cui C, et al.
Stem Cell Research & Therapy, 8(1), 109-109 (2017)

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service