Skip to Content
Merck

SML0279

LG100268

≥98% (HPLC)

Synonym(s):

2-[1-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthyl)cyclopropyl]pyridine-5-carboxylic acid, AGN 192620, ALRT 268., CD 3127, LG268, LGD 100268, LGD 1268

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H29NO2
CAS Number:
Molecular Weight:
363.49
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

Cc1cc2c(cc1C3(CC3)c4ccc(cn4)C(O)=O)C(C)(C)CCC2(C)C

InChI

1S/C24H29NO2/c1-15-12-18-19(23(4,5)9-8-22(18,2)3)13-17(15)24(10-11-24)20-7-6-16(14-25-20)21(26)27/h6-7,12-14H,8-11H2,1-5H3,(H,26,27)

InChI key

SLXTWXQUEZSSTJ-UHFFFAOYSA-N

Biochem/physiol Actions

LG100268 (LG268) is a potent and selective rexinoid and retinoid-X receptor (RXR) agonist. LG100268 binds to the α, β and γ RXR receptors with an IC50 = 3-4 nM and has no activity at the RAR retinoic acid receptors.
LG100268 is a retinoid-X receptor (RXR) agonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library