Skip to Content
Merck
All Photos(2)

Key Documents

B0390

Sigma-Aldrich

Bilirubin Oxidase from Myrothecium verrucaria

greener alternative

lyophilized powder, 15-65 units/mg protein

Synonym(s):

MvBOx, Bilirubin:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Myrothecium verrucaria)

Quality Level

form

lyophilized powder

specific activity

15-65 units/mg protein

composition

Protein, ≥15% biuret

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

toxicology assay: suitable

greener alternative category

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell research. For more information see the article in biofiles.

Bilirubin oxidase (BOD) belongs to the subclass of the multicopper oxidase family. It is usually found in fungi.Bilirubin Oxidase is composed of three cupredoxin-like domains that result in two active sites formed by four copper ions. Research area: Apoptosis

Application

Bilirubin Oxidase from Myrothecium verrucaria has been used to eliminate the interference of bilirubin. It has also been used to chemically modify with fluorescein derivative (FS) and combined with multivariate calibration to analyse direct bilirubin (dBR) and total bilirubin (tBR).
Bilirubin oxidase is used to degrade bilirubin. Bilirubin oxidase, from Myrothecium verrucaria, may be used to determine free hemoglobin in icteric specimens . It also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice.

Biochem/physiol Actions

Bilirubin oxidase (BOD) is a multicopper oxidase that catalyzes the oxidation of bilirubin to biliverdin. Oxygen is the electron acceptor and is reduced to water. The enzyme is used for diagnostic analysis of bilirubin in serum during medical examinations of the serum. It has also been used as an enzymatic catalyst for the cathode of biofuel cells that work under neutral conditions. It is a monomeric protein with a molecular mass of 60 kDa and high reactivity at neutral pH. BOD participates in porphyrin and chlorophyll metabolism.
Bilirubin oxidase (BOD) is capable of oxidizing laccase substrates.

Unit Definition

One unit will oxidize 1.0 μmole of bilirubin per min at pH 8.4 at 37 °C.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laith Hussein et al.
Physical chemistry chemical physics : PCCP, 13(13), 5831-5839 (2011-02-18)
The fabrication process of buckypapers (BPs) made from stable suspensions of as-received or functionalized multi-walled carbon nanotubes (MWCNTs) with high purity (97.5 wt%, Baytubes), their characterization and their utilization towards novel biofuel cell electrode applications are reported. The BPs can
Franziska Wegerich et al.
Langmuir : the ACS journal of surfaces and colloids, 27(7), 4202-4211 (2011-03-16)
Here, we report on cytochrome c/bilirubin oxidase multilayer electrodes with different cytochrome c (cyt c) forms including mutant forms of human cyt c, which exhibit different reaction rates with bilirubin oxidase (BOD) in solution. The multilayer formation via the layer-by-layer
Shannon J Little et al.
Chemical communications (Cambridge, England), 47(31), 8886-8888 (2011-06-16)
A novel bioelectrode system has been prepared in which an enzyme and a conducting polymer hydrogel are combined in a nanostructured scaffold. The latter consists of fibres of carbon NanoWeb, grown by chemical vapour deposition onto reticulated vitreous carbon (RVC).
S S Wong et al.
Annals of clinical and laboratory science, 25(3), 247-251 (1995-05-01)
Owing to the overlapping absorption bands of hemoglobin and bilirubin, spectrophotometric determination of plasma free hemoglobin has been plagued by the interference of bilirubin when the latter is present in significant concentrations. To resolve this problem, bilirubin oxidase from Myrothecium
Akio Ueda et al.
Journal of the American Chemical Society, 133(13), 4840-4846 (2011-03-10)
We have developed a new carbon film electrode material with thornlike surface nanostructures to realize efficient direct electron transfer (DET) with enzymes, which is very important for various enzyme biosensors and for anodes or cathodes used in biofuel cells. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service