Skip to Content
Merck
All Photos(2)

Key Documents

19359

Sigma-Aldrich

Glucose dehydrogenase from Pseudomonas sp.

greener alternative

powder, white, ≥200 U/mg

Synonym(s):

GDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Pseudomonas spp.)

form

powder

specific activity

≥200 U/mg

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white

greener alternative category

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell research. For more information see the article in biofiles.

Application

Glucose dehydrogenase from Pseudomonas sp. has been used for the oxidation of glucose as a part of flow-injection analysis detection. It has also been used to regenerate the consumed nicotinamide adenine dinucleotide (NADH) during the removal of pyruvate by lactate dehydrogenase.

Biochem/physiol Actions

In bacteria, the membrane integrated glucose dehydrogenase catalyses the conversion of glucose to gluconic acid. It uses pyrroloquinoline quinone as a coenzyme. The intracellular glucose dehydrogenase lack this cofactor. It is a nicotinamide adenine dinucleotide (NADH) dependent enzyme.

Unit Definition

One unit corresponds to the amount of enzyme which will oxidizes 1 μmole β-D-glucose to D-glucono-δ-lactone per minute at pH 8.0 and 37 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Nadeem Zafar et al.
Analytical chemistry, 84(1), 334-341 (2011-11-19)
A new extracellular flavin adenine dinucleotide (FAD)-dependent glucose dehydrogenase from Glomerella cingulata (GcGDH) was electrochemically studied as a recognition element in glucose biosensors. The redox enzyme was recombinantly produced in Pichia pastoris and homogeneously purified, and its glucose-oxidizing properties on
Jen-Yuan Wang et al.
Bioresource technology, 116, 502-506 (2012-05-01)
A glucose bio-battery prototype independent of oxygen is proposed based on a glucose dehydrogenase (GDH) bioanode and a graphite cathode with an iodide/tri-iodide redox couple. At the bioanode, a NADH electrocatalyst, poly(methylene blue) (PMB), which can be easily grown on
Mátyás Cserháti et al.
Journal of bacteriology, 194(8), 2109-2110 (2012-03-31)
Here we report on the complete genome sequence of Cupriavidus basilensis OR16 NCAIM BO2487. The genome of strain OR16 contains 7,534 putative coding sequences, including a large set of xenobiotics-degrading genes and a unique glucose dehydrogenase gene that is absent
Basic Biotechnology, 370-370 (2006)
Akasit Siriphongphaew et al.
Applied microbiology and biotechnology, 95(2), 357-367 (2012-05-05)
Oxygenases-based Escherichia coli whole-cell biocatalyst can be applied for catalysis of various commercially interesting reactions that are difficult to achieve with traditional chemical catalysts. However, substrates and products of interest are often toxic to E. coli, causing a disruption of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service