Skip to Content
Merck
All Photos(5)

Key Documents

A3631

Sigma-Aldrich

Arachidic acid

≥99%

Synonym(s):

Arachic acid, Arachidinic acid, Eicosanoic acid, Icosanoic acid, NSC 93983

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18COOH
CAS Number:
Molecular Weight:
312.53
Beilstein:
1788211
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

form

crystalline powder

technique(s)

HPLC: suitable

mp

74-76 °C (lit.)

functional group

carboxylic acid

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)

InChI key

VKOBVWXKNCXXDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Arachidic acid has been used as a fatty acid standard to optimize a sensitive high-performauce liquid chromatography (HPLC) method to analyse fatty acid (FA) for the quantification of polyunsaturated FAs (PUFAs) in cell lipid extracts.

Biochem/physiol Actions

Arachidic acid is a saturated fatty acid with a 20 carbon chain. Arachidic acid occurs naturally in fish and vegetable oils. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATTY ACIDS| Properties
Encyclopedia of food sciences and nutrition (2003)
Lipids in blood-brain barrier models in vitro I: thin-layer chromatography and high-performance liquid chromatography for the analysis of lipid classes and long-chain polyunsaturated fatty acids
Kraamer SD, et al.
In Vitro Cellular & Developmental Biology. Animal, 38(10), 557-565 (2002)
Detection of unusual very-long-chain fatty acid and ether lipid derivatives in the fibroblasts and plasma of patients with peroxisomal diseases using liquid chromatography-mass spectrometry
Takashima S, et al.
Molecular Genetics and Metabolism, 120(3), 255-268 (2017)
Ubonvan Termsarasab et al.
Colloids and surfaces. B, Biointerfaces, 109, 280-286 (2013-05-15)
Arachidyl chitosan (chitosan oligosaccharide-arachidic acid; CSOAA)-based self-assembled nanoprobes for magnetic resonance imaging (MRI) of neoplastic lesions was developed and evaluated in vitro. Diethylenetriaminepentaacetic dianhydride (DTPA) was conjugated to chitosan oligosaccharide (CSO) and Gd(3+) was chelated to the resulting ligand. DTPA
Leiliang Zheng et al.
Journal of the American Society for Mass Spectrometry, 19(1), 96-102 (2008-02-23)
Langmuir-Blodgett multilayers of alternating barium arachidate and barium dimyristoyl phosphatidate are characterized by secondary ion mass spectrometry employing a 40 keV buckminsterfullerene (C60) ion source. These films exhibit well-defined structures with minimal chemical mixing between layers, making them an intriguing

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service