Skip to Content
Merck
All Photos(1)

Key Documents

A8311

Sigma-Aldrich

Antifoam 204

aqueous emulsion for bacterial and mammalian systems

Synonym(s):

organic antifoam

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161901
NACRES:
NA.85

Quality Level

form

emulsion (aqueous)

concentration

100% (organic)

density

1.01 g/mL at 25 °C

application(s)

microbiology

storage temp.

room temp

suitability

(Mammalian (suspension))
bacteria (fermentation)

Looking for similar products? Visit Product Comparison Guide

General description

Antifoam 204 is an organic antifoam used to prevent or minimize foaming in both microbial fermentation systems and mammalian suspension cultures. Control of foaming minimizes damage to the cells, resulting in increased protein/antibody production.

Application

Suitable for preventing or minimizing foaming in microbial fermentation systems and mammalian suspension cultures.
Antifoam 204 has been used:
  • in the expression and purification of PUL (PLAP, Ufd3p, and Lub1p) domain
  • in fed-batch fermentation
  • to prevent bubble formation due in aeration in Luria Bertani (LB) culture medium for bacterial growth
  • in the batch fermentation with mineral medium

Components

Antifoam 204 is a 100% active components and a mixture of non-silicone organic defoamers in a polyol dispersion.

Other Notes

Contains 100% active components and is a mixture of non-silicone organic defoamers in a polyol dispersion. Can be sterilized repeatedly.

Preparation Note

For use in microbiological media Sigma recommends a starting concentration of between 0.005% and 0.01%.
The optimal amount of antifoam required for various applications will need to be determined. Antifoam 204 is soluble in methanol, ethanol, toluene, xylene, perchloroethylene, and cold water at temperatures below 15 °C. It is insoluble in warm water and ethylene glycol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yixin Yang et al.
Biomacromolecules, 11(1), 259-268 (2009-12-17)
Biobased omega-carboxy fatty acid monomers 1,18-cis-9-octadecenedioic, 1,22-cis-9-docosenedioic, and 1,18-cis-9,10-epoxy-octadecanedioic acids were synthesized in high conversion yields from oleic, erucic and epoxy stearic acids by whole-cell biotransformations catalyzed by C. tropicalis ATCC20962. Maximum volumetric yields in shake-flasks were 17.3, 14.2, and
Beth Junker
Biotechnology progress, 23(4), 767-784 (2007-06-15)
Key aspects of foaming and its mitigation in fermentation systems are presented. Foam properties and behavior, conditions that affect foaming, and consequences of foaming are discussed, followed by methods to detect and prevent foam, both without and with the use
Bouke Wim de Jong et al.
FEMS yeast research, 16(1), fov105-fov105 (2015-11-23)
Saccharomyces cerevisiae has previously been engineered to become a cell factory for the production of fatty acid ethyl esters (FAEEs), molecules suitable for crude diesel replacement. To find new metabolic engineering targets for the improvement of FAEE cell factories, three
Angelica Rodriguez et al.
Metabolic engineering, 44, 265-272 (2017-11-05)
The development of robust and efficient cell factories requires understanding of the metabolic changes triggered by the production of the targeted compound. Here we aimed to study how production of p-coumaric acid, a precursor of multiple secondary aromatic metabolites, influences
Junpeng Xiao et al.
Methods in molecular biology (Clifton, N.J.), 751, 329-342 (2011-06-16)
Site-specific modification of glycoproteins has wide application in both biochemical and biophysical studies. This method describes the conjugation of synthetic molecules to the N-terminus of a glycoprotein fragment, viz., human immunoglobulin G subclass 1 fragment crystallizable (IgG1 Fc), by native

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service