Skip to Content
Merck
All Photos(1)

Key Documents

A5906

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Synonym(s):

SAM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

sheep

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule)-Peroxidase antibody is specific for mouse IgG.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Horseradish Peroxidase (HRP) is an enzyme that catalyzes the conversion of chromogenic substrates such as o-phenylenediamine (OPD), 4-chloro-1-naphthol 3,3′,5,5′-tetramethylbenzidine (TMB), 3,3′-Diaminobenzidine (DAB) or 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS); chemiluminescent substrates such as CPS-3 (enhanced luminal) and fluorogenic substrates such as Ampliflu Red into detectable chromophores, light-emitters or fluorescers, respectively.
Sheep polyclonal anti-Mouse IgG (whole molecule)–Peroxidase antibody reacts with mouse IgG in vitro and in mouse serum and biological fluids. It does not react with human serum proteins.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)-Peroxidase antibody is suitable for use in peptide arrays (1:500) and western blot. The product can also be used for direct ELISA (1:10,000).
Sheep polyclonal anti-Mouse IgG (whole molecule)–Peroxidase antibody may be used to detect and quantitate the level of IgG in mouse serum and biological fluids via chromogenic, chemoluminescent or fluorogenic immunochemical or immunohistochemical techniques. It may also be used as a secondary antibody in assays that use mouse IgG as the primary antibody. Immunostaining was performed using HRP-conjugated sheep anti-mouse IgG (SAM) diluted in 1% BSA-TBS. HRP was visualized using Na nitroprusside and o-clianisidine (Sigma).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P La et al.
Oncogene, 25(25), 3537-3546 (2006-02-02)
Menin is encoded by the tumor suppressor gene MEN1 that is mutated in patients with an inherited tumor syndrome, multiple endocrine neoplasia type 1 (MEN1). Although menin is a nuclear protein and directly binds to DNA through its nuclear localization
Victor Tapia et al.
Journal of peptide science : an official publication of the European Peptide Society, 14(12), 1309-1314 (2008-09-26)
A high demand of interest concerning binding assays to study the consequences of posttranscriptional phosphorylation may be addressed by peptide array-based methods. A crucial factor for de novo chemical approaches to generate such arrays is the possibility to rationally permutate
Carl Julien et al.
Acta neuropathologica communications, 6(1), 131-131 (2018-12-01)
Exposure to the β-amyloid peptide (Aβ) is toxic to neurons and other cell types, but the mechanism(s) involved are still unresolved. Synthetic Aβ oligomers can induce ion-permeable pores in synthetic membranes, but whether this ability to damage membranes plays a
Pamela S Lagali et al.
Human molecular genetics, 25(21), 4787-4803 (2017-02-09)
ATRX is a chromatin remodeling protein that is mutated in several intellectual disability disorders including alpha-thalassemia/mental retardation, X-linked (ATR-X) syndrome. We previously reported the prevalence of ophthalmological defects in ATR-X syndrome patients, and accordingly we find morphological and functional visual
Orsolya I Hoffmann et al.
PloS one, 11(5), e0154489-e0154489 (2016-05-07)
The Sleeping Beauty transposon system was established as a robust and efficient method for germline transgenesis in different mammalian species. The generation of transgenic mice, rats, rabbits and swine carrying an identical Venus reporter construct delivered by transposon-mediated gene transfer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service