Skip to Content
Merck
All Photos(3)

Key Documents

A4669

Sigma-Aldrich

Acycloguanosine

≥99% (HPLC), powder

Synonym(s):

9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (HPLC)

form

powder

color

white

solubility

H2O: 0.7 mg/mL
1 M HCl: 50 mg/mL
DMSO: 7 mg/mL

ε (extinction coefficient)

11.8 at 256 nm at 1 mM

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes

originator

GlaxoSmithKline

SMILES string

NC1=Nc2c(ncn2COCCO)C(=O)N1

InChI

1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)

InChI key

MKUXAQIIEYXACX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Acycloguanosine or acyclovir is a guanosine analog.

Application

Acycloguanosine has been used as an inhibitor of herpes simplex virus type I:
  • to serve as a positive control to compare the antiviral activities of mushroom extracts in cytotoxic assays using Vero cells
  • in infection studies to test its effect on voltage-gated sodium channels (VGSC) using human dorsal root ganglion-derived neuronal (HD10.6) cells
  • to test its effect on the interferon-stimulated gene (ISG) expression induction (Mx1 and ISG15) in human foreskin fibroblast cells

Biochem/physiol Actions

Acycloguanosine is an antiviral agent and is converted to acycloguanosine triphosphate by herpes simplex virus thymidine kinase (HSV-TK). It competitively inhibits the viral DNA polymerase. It is less effective against cytomegalovirus and Epstein-Barr virus. Acycloguanosine has been used to study herpes simplex virus latency. It may act against human immunodeficiency virus 1 (HIV-1) as well.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faith O Osinaga et al.
Pharmaceuticals (Basel, Switzerland), 16(8) (2023-08-26)
We reported that gamma-hydroxybutyrate (GHB) is released upon Herpes Simplex Virus Type-1 (HSV-1) acute infection. However, the cellular biochemical processes involved in the production of GHB in infected cells are unclear. This study aims to shed light on the biochemical
Haniza Hassan et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-13)
Acyclovir is an antiviral drug used for the treatment of herpes simplex virus infection. Its oral bioavailability is low; therefore, frequent and high doses are prescribed for optimum therapeutic efficacy. Moreover, the current therapeutic regimen of acyclovir is associated with
Antiviral nanodelivery systems: current trends in acyclovir administration
Haniza H, et al.
Journal of Nanomaterials (2016)
J J O'Brien et al.
Drugs, 37(3), 233-309 (1989-03-01)
Acyclovir (aciclovir) is a nucleoside antiviral drug with antiviral activity in vitro against members of the herpes group of DNA viruses. As an established treatment of herpes simplex infection, intravenous, oral and to a lesser extent topical formulations of acyclovir
Hasan Hüseyin Doğan et al.
International journal of medicinal mushrooms, 20(3), 201-212 (2018-05-03)
Despite considerable recent work to reveal different features of mushrooms species, the few studies of antiviral activities are inadequate and therefore further studies are required. Morchella conica, M. esculenta, Terfezia boudieri, Pleurotus ostreatus, Tricholoma anatolicum, Fomes fomentarius, Laetiporus sulphureus, Phellinus

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service