Skip to Content
Merck
All Photos(1)

Key Documents

45396

Supelco

Chlorpyrifos-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7Cl3NO3PS
CAS Number:
Molecular Weight:
322.53
Beilstein:
1541078
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C7H7Cl3NO3PS/c1-12-15(16,13-2)14-7-5(9)3-4(8)6(10)11-7/h3H,1-2H3

InChI key

HRBKVYFZANMGRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Jun-Ran Kim et al.
Biodegradation, 20(4), 487-497 (2008-12-17)
A chlorpyrifos-methyl (CM) degrading bacterium (designated strain KR100) was isolated from a Korean rice paddy soil and was further tested for its sensitivity against eight commercial antibiotics. Based on morphological, biochemical, and molecular characteristics, this bacterium showed greatest similarity to
Tong Wu et al.
Environmental science & technology, 40(3), 784-790 (2006-03-03)
Chlorpyrifos-methyl is widely used in the control of insects on certain stored grain, including wheat, barley, oats, rice, and sorghum. The reactions of chlorpyrifos-methyl with hydrogensulfide/bisulfide (H2S/HS-), polysulfides (Sn(2-)), thiophenolate (PhS-), and thiosulfate (S2O3(2-)) were examined in well-defined aqueous solutions
Amalia Muñoz et al.
Environmental science & technology, 45(5), 1880-1886 (2011-02-04)
The gas-phase atmospheric degradation of chlorpyrifos-methyl (a widely used organophosphate insecticide in Southern European regions) has been investigated at the large outdoor European Photoreactor (EUPHORE) in Valencia, Spain. Photolysis under sunlight conditions and reaction with ozone were shown to be
M Chouaïbou et al.
Tropical medicine & international health : TM & IH, 13(4), 476-486 (2008-02-06)
To explore temporal variation in insecticide susceptibility of Anopheles gambiae s.l. populations to the four chemical groups of insecticides used in public health and agriculture, in close match with the large-scale cotton spraying programme implemented in the cotton-growing area of
Raphael N'Guessan et al.
Malaria journal, 9, 44-44 (2010-02-10)
Scaling up of long-lasting insecticidal nets (LLINs) and indoor residual spraying (IRS) with support from the Global Fund and President's Malaria Initiative is providing increased opportunities for malaria control in Africa. The most cost-effective and longest-lasting residual insecticide DDT is

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service