Skip to Content
Merck

Skip To

A4327

Albumin from human serum

lyophilized powder, essentially protease free, ≥96% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
EC Number:
274-272-6
MDL number:
eCl@ss:
42010201
Biological source:
human
Assay:
≥96% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
tissue culture: suitable
Impurities:
HIV I and HIVII, HCV and HBsAg, tested negative
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

tissue culture: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

foreign activity

Alkaline phosphatase ≤0.001 units/mg protein, Peroxidase ≤0.001 units/mg protein, Protease ≤0.0001 units/mg protein

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Albumin is the most copious protein in blood plasma. Produced in the liver,[1] serum albumin serves as a carrier protein for biomolecules like as fatty acids, amino acids, and steroids.

This human serum albumin (HSA) product has been specially tested and evaluated to be low in the following properties:
  • Essentially protease-free / low protease content
  • Low alkaline phosphatase content
  • Low peroxidase content

Serum albumin is used in various applications related to molecular biology, notably as an inert protein stabilizer for stock solutions of enzymes and proteins of interest. In such applications, it is thus important for the HSA to be protease free, to maintain the integrity of the protein or enzyme of interest.

Protease-free HSA is especially useful for applications like:
  • Enzyme assays
  • Protein-based assays
  • Protease-sensitive techniques such as:
(a) Enzyme immunoassay (EIA)
(b) Nucleic acid hybridization
(c) Radioimmunoassay (RIA)

Application

Albumin was used to test its effect on the in vitro bactericidal activity of cefditoren against penicillin-resistant Streptococcus pneumonia.[2] It has been clinically used in serious and often life-threatening conditions, such as shock and blood loss due to trauma, burns, and surgery.[1]
It was used to test the effect of non-enzymatic glycation on the unfolding of human serum albumin.[3] It was also used to test the two distinct pathways used by intestinal epithelial cells for HLA class II antigen processing.[4]

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Features and Benefits

  • Low protease activity / essentially protease-free (≤ 0.0001 units/mg protein)
  • Low alkaline phosphatase activity (≤ 0.001 units/mg protein)
  • Low peroxidase activity (≤ 0.001 units/mg protein)

Preparation Note

Prepared from A1653

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A1653A8763A9080
assay

≥96% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

-

biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

tissue culture: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

liquid

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)


Still not finding the right product?

Explore all of our products under Albumin from human serum


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters