79033
(R)-(+)-1,2-Octanediol
≥95.0% (sum of enantiomers, GC)
Synonym(s):
Octane-1,2-diol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)5CH(OH)CH2OH
CAS Number:
Molecular Weight:
146.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥95.0% (sum of enantiomers, GC)
form
flakes
SMILES string
CCCCCC[C@@H](O)CO
InChI
1S/C8H18O2/c1-2-3-4-5-6-8(10)7-9/h8-10H,2-7H2,1H3/t8-/m1/s1
InChI key
AEIJTFQOBWATKX-MRVPVSSYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
suitable for GCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service