Skip to Content
Merck
All Photos(2)

Key Documents

33393

Supelco

2,3,4-Trichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
1945113
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

75-79 °C (lit.)
75-79 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccc(Cl)c(Cl)c1Cl

InChI

1S/C6H3Cl3O/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H

InChI key

HSQFVBWFPBKHEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4-Trichlorophenol may be used as a reference standard for the determination of the analyte in water samples using gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D de Almeida Azevedo et al.
Journal of chromatography. A, 879(1), 13-26 (2000-06-28)
Gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS) were optimized and applied for the trace-level determination of 42 priority pesticides and 33 priority organic pollutants from European Union Directive EC 76/464. First, off-line solid-phase extraction
Yanjun Fang et al.
Journal of proteomics, 110, 172-182 (2014-08-12)
Pentachlorophenol (PCP) and 2,4,6-trichlorophenol (TCP) are suspected of disrupting the endocrine system and thus affecting human and wildlife reproduction, but the potential common mechanisms and biomarkers of chlorophenols (CPs) in the ovary are not fully elucidated. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service