Skip to Content
Merck
All Photos(1)

Documents

74250

Supelco

Nonane

analytical standard

Synonym(s):

n-Nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH3
CAS Number:
Molecular Weight:
128.26
Beilstein:
1696917
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.41 (vs air)

vapor pressure

0.18 psi ( 37.7 °C)

Assay

≥99.8% (GC)

autoignition temp.

401 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.405 (lit.)
n20/D 1.405

bp

151 °C (lit.)

mp

−53 °C (lit.)

density

0.718 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCC

InChI

1S/C9H20/c1-3-5-7-9-8-6-4-2/h3-9H2,1-2H3

InChI key

BKIMMITUMNQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Faist et al.
European journal of medicinal chemistry, 45(1), 179-185 (2009-11-03)
N-Alkyl and N-(2-dialkylaminoethyl) derivatives of 5-amino-2-azabicyclo-nonanes were prepared and tested in vitro for their activities against the multidrug-resistant K1 strain of Plasmodium falciparum and Trypanosoma brucei rhodesiense (STIB 900). Most of the new compounds showed lower antitrypanosomal activity than their
Jeongmi Choi et al.
Journal of chromatography. A, 1218(41), 7227-7233 (2011-09-13)
In order to analyze amino acids sensitively without derivatization, we have developed carrier-mediated single drop microextraction (SDME). Nonane-1-sulfonic acid was added to an acidic sample donor solution as a carrier to form neutral ion pair complexes with amino acids. The
Daniel H Ess et al.
Organic letters, 11(23), 5538-5541 (2009-11-11)
Density functional theory was used to locate transition states for hydroboration reactions of allenes with 9-borabicyclo[3.3.1]nonane and 10-R-9-borabicyclo[3.3.2]decane, as well as transition states for [1,3]-boratropic shift and aldehyde addition reactions of the derived allylboranes. The origin of kinetic versus thermodynamic
Arif Baran et al.
The Journal of organic chemistry, 77(3), 1244-1250 (2012-01-11)
1,3,3a,7a-Tetrahydro-2-benzofuran was used as key compound for the synthesis of various bishomoinositol derivatives. The diene was subjected to an epoxidation reaction for further functionalization of the diene unit. The bisepoxide obtained was submitted to a ring-opening reaction with acid in
Arif Baran et al.
The Journal of organic chemistry, 77(11), 5086-5097 (2012-05-23)
Transformation of cyclohexa-2,4-diene-1,2-diylbis(methylene) diacetate to various carbasugars is described. Photooxygenation of a cyclohexadiene derivative gave a bicyclicendoperoxide, which was reduced with thiourea to [2-[(acetyloxy)methyl]cyclohexa-2,4-dien-1-yl]methyl acetate. Epoxidation of the remaining double bond followed by epoxide ring-opening and hydrolysis of the acetate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service