Skip to Content
Merck
All Photos(1)

Key Documents

71578

Sigma-Aldrich

Poly(ethylene glycol) methyl ether

BioUltra, 500

Synonym(s):

Polyethylene glycol, Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.26

vapor density

>1 (vs air)

vapor pressure

0.05 mmHg ( 20 °C)

description

non-ionic

product line

BioUltra

form

paste
viscous liquid (clear)

mol wt

470-530

impurities

insoluble matters, passes filter test
≤0.005% peroxides (as H2O2)

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

20 °C (lit.)

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.03

functional group

hydroxyl

SMILES string

O(CCO)C

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wei et al.
Langmuir : the ACS journal of surfaces and colloids, 28(39), 13984-13992 (2012-09-04)
The microcosmic mechanisms of protein (recombinant human growth hormone, rhGH) incomplete release and stability from amphiphilic poly(monomethoxypolyethylene glycol-co-D,L-lactide) (mPEG-PLA, PELA) microspheres were investigated. PELA with different hydrophilicities (PELA-1, PELA-2, and PELA-3) based on various ratios of mPEG to PLA were
Jiani Zheng et al.
Langmuir : the ACS journal of surfaces and colloids, 28(37), 13261-13273 (2012-08-28)
Alginate/chitosan/alginate (ACA) hydrogel microcapsules were modified with methoxy poly(ethylene glycol) (MPEG) to improve protein repellency and biocompatibility. Increased MPEG surface graft density (n(S)) on hydrogel microcapsules was achieved by controlling the grafting parameters including the buffer layer substrate, membrane thickness
Lei Liu et al.
International journal of pharmaceutics, 443(1-2), 175-182 (2013-01-05)
This work aims to develop curcumin (Cur) loaded biodegradable self-assembled polymeric micelles (Cur-M) to overcome poor water solubility of Cur and to meet the requirement of intravenous administration. Cur-M were prepared by solid dispersion method, which was simple and easy
Yiyi Yu et al.
Journal of pharmaceutical sciences, 102(3), 1054-1062 (2013-01-03)
To promote the application of methoxy poly(ethylene glycol)-cholesterol (mPEG-Chol), mPEG-Chol was used to prepare core-shell micelles encapsulating poorly water-soluble docetaxel (DTX-PM) by modified cosolvent evaporation method. Approaches to enhance DTX entrapment efficiency (EE) and minimize particle size were investigated in
Pengxiang Zhao et al.
Chemical communications (Cambridge, England), 49(31), 3218-3220 (2013-03-14)
"Click" chemistry now offers access to a great variety of triazoles, and the first example of a strategy to stabilize gold nanoparticles (AuNPs) with a new 1,2,3-triazole-mPEG ligand is developed here together with preliminary examples of possible applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service