Skip to Content
Merck
All Photos(2)

Key Documents

202398

Sigma-Aldrich

Poly(ethylene glycol)

greener alternative

average Mn 400, hydroxyl

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol), average Mn 400

form

viscous liquid

mol wt

Mn 380-420
average Mn 400

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.466

pH

4.5-7.5

viscosity

7.3 cSt(210 °F)(lit.)

mp

4-8 °C (lit.)

density

1.128 g/mL

Ω-end

hydroxyl

α-end

hydroxyl

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a synthetic, hydrophilic, biocompatible polymer with widespread use in biomedical and other applications. PEGs are non-toxic, FDA-approved, generally non-immunogenic, and frequently used in many biomedical applications including bioconjugation, drug delivery, surface functionalization, and tissue engineering. In drug delivery, PEGs can be used as linkers for antibody-drug conjugates (ADCs) or as a surface coating on nanoparticles to improve systemic drug delivery.

Application

Poly(ethylene glycol) (PEG) has been used as a coating agent for the synthesis and in vitro safety assessment of magnetic bacterial cellulose (MBC) pellicles, performed on porcine aortic smooth muscle cells (PAOSMC). PEG provides a compatible surface for cell culture, since uncoated iron oxide NPs are considered cytotoxic to cells.[1] It has also been used as a plasticizer in investigating the effectiveness of polyvinylpyrrolidone microneedles for transdermal delivery of functional collagen using porcine skin, adult human skin analog and human foreskin. Microneedles obtained, were reportedly characterized by various analytical and bio-analytical techniques.[2]
Used in conjunction with carbon black to form a conductive composite.[3] Polymer nanospheres of poly(ethylene glycol) were used for drug delivery.[4]

Legal Information

Selectfluor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transdermal delivery of functional collagen via polyvinylpyrrolidone microneedles
Sun W, et al.
Annals of Biomedical Engineering, 43(12), 2978-2990 (2015)
Changxin Wan et al.
Cancer research, 81(1), 158-173 (2020-11-08)
Immune therapies have had limited efficacy in high-grade serous ovarian cancer (HGSC), as the cellular targets and mechanism(s) of action of these agents in HGSC are unknown. Here we performed immune functional and single-cell RNA sequencing transcriptional profiling on novel
Ruth Hernandez-Perez et al.
Physical review. E, 96(6-1), 062803-062803 (2018-01-20)
The evaporation of droplets can give rise to a wide range of interesting phenomena in which the dynamics of the evaporation are crucial. In this work, we find simple scaling laws for the evaporation dynamics of axisymmetric droplets pinned on
Synthesis and in vitro safety assessment of magnetic bacterial cellulose with porcine aortic smooth muscle cells
Pastrana FH, et al.
Journal of Biomedical Materials Research Part A, 104(11), 2801-2809 (2016)
Akashi, M., Akagi, T.
Yuki Gosei Kaguku Kyokaishi, 65, 520-520 (2004)

Related Content

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Questions

1–3 of 3 Questions  
  1. Hi, I wanted to know how did you prepare the PEG 400 into liquid form as it in my concern it might not be 100 % PEG would it be like 90% of PEG powdered form in aqueou solution or so on. I will be thankful if you provide answer of my this query.

    1 answer
    1. The specific manufacturing process for the product is proprietary. PEG400 is a pure liquid form of polyethylene glycol with no other components, and its viscous nature is a characteristic feature of the product.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service