Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
445878
Poly(ethylene glycol) dimethyl ether
average MN 250, cross-linking reagent click chemistry
Synonym(s):
Polyethylene glycol, Dimethyl polyethylene glycol
About This Item
Recommended Products
Product Name
Poly(ethylene glycol) dimethyl ether, average Mn ~250
form
liquid
mol wt
average Mn ~250
reaction suitability
reagent type: cross-linking reagent
reaction type: click chemistry
refractive index
n20/D 1.441
bp
>250 °C
density
1.03 g/mL at 25 °C
polymer architecture
shape: linear
functionality: homobifunctional
SMILES string
CO.OCCO
InChI
1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3
InChI key
XTHFKEDIFFGKHM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Paper Title: Molecular dynamics simulations for self-diffusion coefficients: The case of CO2, n-alkanes, and poly(ethylene glycol) dimethyl ethers - Studies self-diffusion coefficients using molecular dynamics simulations to provide insights into the behavior of poly(ethylene glycol) dimethyl ethers (OA Moultos et al., 2016).
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
312.8 °F - closed cup
Flash Point(C)
156.00 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
What is the viscosity of this material at room temperature?
1 answer-
The viscosity is not tested (and therefore, guaranteed) for this material, but there is another product with the same average molecular weight offered (https://www.sigmaaldrich.com/product/mm/814173), which does have a viscosity value listed, which is 7.8 cSt (20 °C). The viscosity of product 445878 should be similar.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service