Skip to Content
Merck
All Photos(3)

Key Documents

175366

Sigma-Aldrich

Stearic acid

Stearic acid

reagent grade, 95%

Synonym(s):

STA, 1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor pressure

1 mmHg ( 173.7 °C)

Assay

95%

form

powder or flakes

bp

361 °C (lit.)

mp

67-72 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Stearic acid (SA) is a straight chain fatty acid. It forms ordered monolayered Langmuir-Blodgett (LB) films on the surface of germanium. These LB films have been investigated by Fourier transform-infrared (FTIR)-attenuated total reflection (AR) spectra.[1]

Conversion of SA with >98% selectivity to deoxygenated C17 products by employing carbon supported palladium catalyst has been reported.[2] It is a promising phase change material (PCM) due to its non-toxicity and convenient melting temperature. Also, it exhibits almost negligible supercooling while undergoing phase change.[3]

Application

Stearic acid can undergo esterification in the presence of mesoporous solid acid catalysts to form methyl stearate.[4] Due to its waterproof ability, STA may be used to impart superhydrophobicity on various surfaces.[5] STA may also be used in the preparation of stearic acid self-assembled monolayers.[6]
Stearic acid may be used to compose the core during sol-gel synthesis of nanoencapsulated phase change materials (NEPCMs).[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

392.0 °F - Cleveland open cup

Flash Point(C)

200 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis and thermal performances of stearic acid/titania core/shell nanocapsules by sol-gel method.
Latibari ST, et al.
Energy, 85, 635-644 (2015)
Heterogeneous catalytic deoxygenation of stearic acid for production of biodiesel.
Latibari ST, et al.
Industrial & Engineering Chemistry Research, 45(16), 5708-5715 (2006)
Nano-crystalline, mesoporous aerogel sulfated zirconia as an efficient catalyst for esterification of stearic acid with methanol.
Saravanan K, et al.
Applied Catalysis. B, Environmental, 192, 161-170 (2016)
FTIR-ATR studies on Langmuir-Blodgett films of stearic acid with 1-9 monolayers.
Kimura F, et al.
Langmuir, 2(1), 96-101 (1986)
Study on superhydrophobic hybrids fabricated from multiwalled carbon nanotubes and stearic acid.
Wu T, et al.
Journal of Colloid and Interface Science, 348(1), 265-270 (2010)

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Solublity of this product?

    1 answer
    1. This product is not tested for solubility. However, other Stearic acid products available are tested for solubility in chloroform at 100 mg/mL. Other sources report that this compound is also soluble in several other solvents:
      acetone: soluble
      benzene: freely soluble
      ethanol: soluble
      ether: freely soluble
      propylene glycol: soluble

      Helpful?

  4. I am curious about the storage temperature for this product. Can you tell me?

    1 answer
    1. This product is stored at ambient temperature. If no storage temperature is listed in the PROPERTIES section of the product page or the product label it may be assumed that ambient temperature is suitable.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service