Skip to Content
Merck
All Photos(1)

Key Documents

67563

Sigma-Aldrich

3,4-Dihydroxy-5-methoxycinnamic acid

≥95.0% (HPLC)

Synonym(s):

3-(3,4-Dihydroxy-5-methoxyphenyl)-2-propenoic acid, 3-Methoxycaffeic acid, 5-Hydroxyferulic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O5
CAS Number:
Molecular Weight:
210.18
Beilstein:
2697317
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (HPLC)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COc1cc(\C=C\C(O)=O)cc(O)c1O

InChI

1S/C10H10O5/c1-15-8-5-6(2-3-9(12)13)4-7(11)10(8)14/h2-5,11,14H,1H3,(H,12,13)/b3-2+

InChI key

YFXWTVLDSKSYLW-NSCUHMNNSA-N

Biochem/physiol Actions

Secondary plant metabolite of the phenylpropanoid pathway.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Maury et al.
Plant physiology, 121(1), 215-224 (1999-09-11)
The biosynthesis of lignin monomers involves two methylation steps catalyzed by orthodiphenol-O-methyltransferases: caffeic acid/5-hydroxyferulic acid 3/5-O-methyltransferases (COMTs) and caffeoyl-coenzyme A (CoA)/5-hydroxyferuloyl-CoA 3/5-O-methyltransferases (CCoAOMTs). Two COMT classes (I and II) were already known to occur in tobacco (Nicotiana tabacum) and three
C C Chapple et al.
The Plant cell, 4(11), 1413-1424 (1992-11-01)
Mutants of Arabidopsis deficient in a major leaf phenylpropanoid ester, 2-O-sinapoyl-L-malate, were identified by thin-layer chromatographic screening of methanolic leaf extracts from several thousand mutagenized plants. Mutations at a locus designated SIN1 also eliminate accumulation of the sinapic acid esters
John M Humphreys et al.
Current opinion in plant biology, 5(3), 224-229 (2002-04-19)
Considerable interest in lignin biosynthesis has been fueled by the many roles that lignin plays in development and in resistance to biotic and abiotic stress, as well as its importance to industry and agriculture. Although the pathway leading to the
K Inoue et al.
Archives of biochemistry and biophysics, 375(1), 175-182 (2000-02-23)
Caffeic acid/5-hydroxyferulic acid 3/5-O-methyltransferase (COMT, EC 1.2.1.68) catalyzes at least two reactions in lignin biosynthesis. Of its two supposed substrates in the lignin pathway, COMT from most sources methylates 5-hydroxyferulic acid (5HFA) with two to three times higher activity than
Probing the mysteries of lignin biosynthesis: the crystal structure of caffeic acid/5-hydroxyferulic acid 3/5-O-methyltransferase provides new insights.
Nancy A Eckardt
The Plant cell, 14(6), 1185-1189 (2002-06-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service