Skip to Content
Merck
All Photos(3)

Key Documents

D7927

Sigma-Aldrich

Sinapic acid

≥98%, powder

Synonym(s):

3,5-Dimethoxy-4-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxy-cinnamic acid, Sinapinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O5
CAS Number:
Molecular Weight:
224.21
Beilstein:
2699118
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mol wt

monoisotopic mol wt 224.06839 Da

mp

~202 °C

solubility

H2O: slightly soluble (lit.)(lit.)
methanol: water: soluble (lit.)(lit.)
polar organic solvents: soluble (lit.)(lit.)

SMILES string

COc1cc(\C=C\C(O)=O)cc(OC)c1O

InChI

1S/C11H12O5/c1-15-8-5-7(3-4-10(12)13)6-9(16-2)11(8)14/h3-6,14H,1-2H3,(H,12,13)/b4-3+

InChI key

PCMORTLOPMLEFB-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sinapic acid is an hydroxycinnamic acid derivative that occurs naturally in Brassicaceae species .

Sinapic acid is a phenylpropanoid derivative that acts as a radical scavenger with antioxidant, antimicrobial, anti-inflammatory, and anticancer properties.

Application

Sinapic acid can be used:
  • As a potent peroxynitrite (ONOO−) oxidant scavenger for the protection of the cellular constituents against peroxynitrite cytotoxic species.
  • In the synthesis of antioxidant hydroxycinnamic acid xylan esters via esterification reaction.
  • In the synthesis of pseudo-cinnamic derivatives effective against mycobacterium tuberculosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Feng Huang et al.
Journal of inflammation research, 13, 883-895 (2020-11-20)
Low back pain (LBP) is a very common condition and leads to serious pain, disability, and price tag all over the world. Intervertebral disk degeneration (IDD) is one of the major reasons that contributed to LBP. The levels of interleukin
Sinapic acid: is it safe for humans?
Hasan H et al.
FABAD Journal of Pharmaceutical Sciences, 41, 39-39 (2016)
Sinapic acid and its derivatives: natural sources and bioactivity.
Niciforovic N and Abramovivc H
Comprehensive Reviews in Food Science and Food Safety, 13(1), 34-51 (2014)
Peroxynitrite scavenging activity of sinapic acid (3, 5-dimethoxy-4-hydroxycinnamic acid) isolated from Brassica juncea.
Zou Y, et al.
Journal of Agricultural and Food Chemistry, 50(21), 5884-5890 (2002)
Synthesis and antioxidant activity of hydroxycinnamic acid xylan esters.
Wrigstedt P, et al.
Journal of Agricultural and Food Chemistry, 58(11), 6937-6943 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service