Skip to Content
Merck
All Photos(3)

Key Documents

382051

Sigma-Aldrich

4-Hydroxy-3-methoxycinnamaldehyde

98%

Synonym(s):

Coniferyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH=CHCHO
CAS Number:
Molecular Weight:
178.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

175 °C/5 mmHg (lit.)

mp

80-82 °C (lit.)

functional group

aldehyde

SMILES string

COc1cc(\C=C\C=O)ccc1O

InChI

1S/C10H10O3/c1-13-10-7-8(3-2-6-11)4-5-9(10)12/h2-7,12H,1H3/b3-2+

InChI key

DKZBBWMURDFHNE-NSCUHMNNSA-N

General description

4-Hydroxy-3-methoxycinnamaldehyde is an isoflavonoid. It has been isolated from the insecticidally active hot dichloromethane extract of heartwood of Gliricidia sepium, seed kernels of Melia azedarach L and barks of Cinnamomum cebuense.

Application

4-Hydroxy-3-methoxycinnamaldehyde is suitable for use in a study to investigate the antioxidant and antiradical activities of ferulates using a β-carotene-linoleate model system and a DPPH radical scavenging assay, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Isoflavonoids and a pterocarpan from< i> Gliricidia sepium</i>.
Herath HMTB, et al.
Phytochemistry, 47(1), 117-119 (1998)
M Lisa Manier et al.
Journal of the American Society for Mass Spectrometry, 22(8), 1409-1419 (2011-09-29)
Isoniazid (INH) is an important component of front-line anti-tuberculosis therapy with good serum pharmacokinetics but unknown ability to penetrate tuberculous lesions. However, endogenous background interferences hinder our ability to directly analyze INH in tissues. Chemical derivatization has been successfully used
María C Carpinella et al.
Journal of agricultural and food chemistry, 53(8), 2922-2927 (2005-04-14)
In the continuous search for antifungal compounds from plants, the hydroxycoumarin scopoletin (1) was isolated from seed kernels of Melia azedarach L. from which three other compounds, vanillin (2), 4-hydroxy-3-methoxycinnamaldehyde (3), and (+/-) pinoresinol (4), have also been isolated. Guided
Antioxidant and antiradical activity of ferulates.
Karamac M, et al.
Czech Journal of Food Sciences, 23(2), 469-475 (2005)
Consolacion Y Ragasa et al.
Chinese journal of natural medicines, 11(3), 264-268 (2013-06-04)
To investigate the chemical constituents of Cinnamomum cebuense, an endemic and critically endangered tree found only in Cebu, Philippines. The compounds were isolated by silica gel chromatography. The structures of the isolates were elucidated by NMR spectroscopy. The dichloromethane (DCM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service