Skip to Content
Merck
All Photos(1)

Key Documents

46290

Sigma-Aldrich

Sudan Red 7B

for microscopy (Bot., Hist.)

Synonym(s):

Fat Red 7B, Fat red bluish, Solvent Red 19

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21N5
CAS Number:
Molecular Weight:
379.46
Colour Index Number:
26050
Beilstein:
3534323
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bot., Hist.)

form

powder

mp

130 °C (dec.) (lit.)

solubility

acetone: 0.1 g/10 mL, clear to turbid, red

λmax

367-377 nm in toluene (2nd)
528-538 nm in toluene (1st)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCNc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C24H21N5/c1-2-25-23-17-12-18-8-6-7-11-22(18)24(23)29-28-21-15-13-20(14-16-21)27-26-19-9-4-3-5-10-19/h3-17,25H,2H2,1H3/b27-26+,29-28+

InChI key

VKWNTWQXVLKCSG-ZDXBJJIESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sudan red 7B, also known as fat red, is a non-fluorescent stain. It is effectively used to stain lipids and is used to indicate the presence of suberin in cell walls. Sudan red 7B acts as a marker for formosan subterranean termite, Coptotermes fornosanus Shiraki.
Appl- Sudan Red 7B has been used to stain root sections of pearl millet.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Root Methods: A Handbook, 594-594 (2013)
Component traits of plant water use are modulated by vapour pressure deficit in pearl millet (Pennisetum glaucum (L.) R. Br.).
Kholova J, et al.
Functional plant biology, 43(5), 423-437 (2016)
Sudan Red 7B, a Dye Marker for Coptotermes formosanus
Lai P Y, et al.
Hawaiian Entomological Society. (1983)
Effects of a dye, Sudan Red 7B, on the Formosan subterranean termite, Coptotermes formosanus Shiraki (Isoptera: Rhinotermitidae).
Su N Y, et al.
Material and Organismen, 18, 127-133 (1983)
Onions and Other Vegetable Alliums, 454-454 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service