Skip to Content
Merck
All Photos(3)

Key Documents

S4131

Sigma-Aldrich

Sudan III

Dye content, ≥85%, technical grade, powder

Synonym(s):

1-[4-(Phenylazo)phenylazo]-2-naphthol, Cerasin Red, Fat Ponceau G, Fat Soluble Sudan, Scarlet B, Solvent Red 23, Sudan G, Sudan Red BK, Tony Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O
CAS Number:
Molecular Weight:
352.39
Colour Index Number:
26100
Beilstein:
931185
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Sudan III, Technical grade

grade

technical grade

Quality Level

form

powder

composition

Dye content, ≥85%

mp

199 °C (dec.) (lit.)

solubility

chloroform: 1 mg/mL, clear, red

λmax

508-512 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/b24-23+,26-25+

InChI key

FHNINJWBTRXEBC-QSZPNPOGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sudan III is a non-ionic lipophilic dye. It belongs to the class of azo dyes. Sudan III is used for tissue ceroid and lipofuscin analysis and blood-cell staining. It is used to color waxes, oils and spirit varnishes. Sudan III stains triglycerides in frozen sections.

Application

Sudan III has been used as an absorber to make turbid phantoms. It has also been used as a colorant to determine emulsification capacity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.L.F. Armarego
Purification of Laboratory Chemicals (2017)
CYP-450 isoenzymes catalyze the generation of hazardous aromatic amines after reaction with the azo dye Sudan III
Food And Chemical Toxicology (2013)
Preventive Effects of Puerarin on Alcohol-induced Osteonecrosis
Clinical Orthopaedics and Related Research (2008)
Analytical and experimental aspects of optical property determination of turbid media using frequency-domain ptr
Analytical Sciences/Supplements Proceedings of 11th International Conference of Photoacoustic and Photothermal Phenomena, s326-s329 (2002)
Rosana Aparecida da Silva-Buzanello et al.
Asian-Australasian journal of animal sciences, 32(8), 1186-1194 (2019-02-13)
The influence of broiler carcass scalding conditions on chicken breast meat quality parameters was investigated. Two hundred and seventy Cobb broiler chickens from 42 to 48 days old were slaughtered according to the standard industry practice and scalded in five

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service