Skip to Content
Merck

Skip To

41707

2-C-Methyl-D-erythritol

≥90% (GC)

Synonym(s):

(2S,3R)-2-Methyl-1,2,3,4-tetrahydroxybutane, (2S,3R)-2-Methylbutane-1,2,3,4-tetrol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H12O4
CAS Number:
Molecular Weight:
136.15
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5916602
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Carum ajowan fruit)

Quality Segment

assay

≥90% (GC)

form

viscous liquid (clear)

color

colorless

storage temp.

2-8°C

SMILES string

C[C@](O)(CO)[C@H](O)CO

InChI

1S/C5H12O4/c1-5(9,3-7)4(8)2-6/h4,6-9H,2-3H2,1H3/t4-,5+/m1/s1

InChI key

HGVJFBSSLICXEM-UHNVWZDZSA-N

General description

(2S,3R)-2-methylbutane-1,2,3,4-tetrol was identified as a glucide in methanol extracts of Carum ajowan fruit.

Biochem/physiol Actions

Metabolite of the non-mevalonate MEP pathway, generally found in prokaryotes, as precursor to isoprenoids as well as non-isoprenoids like vitamins. As this pathway is not present in humans, it is of interest for the development of bacterium-specific drugs in the search for treatments of infectious diseases.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E7500E7625PHR1479
biological source

(Carum ajowan fruit)

biological source

-

biological source

-

biological source

-

assay

≥90% (GC)

assay

≥99% (GC)

assay

≥75% (TLC)

assay

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300

form

viscous liquid (clear)

form

powder

form

syrup

form

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

color

colorless

color

white

color

colorless to very dark yellow

color

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters