Skip to Content
Merck
All Photos(2)

Key Documents

E7500

Sigma-Aldrich

meso-Erythritol

≥99% (GC)

Synonym(s):

1,2,3,4-Butanetetrol, meso-1,2,3,4-Tetrahydroxybutane, i-Erythritol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
Beilstein:
1719753
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (GC)

form

powder

sweetness

0.7 × sucrose

color

white

bp

329-331 °C (lit.)

mp

118-120 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC[C@@H](O)[C@@H](O)CO

InChI

1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+

InChI key

UNXHWFMMPAWVPI-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meso-Erythritol (i-Erythritol) is a precursor to the important tanning agent L-erythrulose which can be commercially generated by Gluconobacter oxydans DSM 7145. Meso-Erythritol may be used to support the carbon source growth requirement of Gluconobacter bacteria. It is an aliphatic poly-alcohol used in the chemical analysis of the reactivity of various radicals and radical anions. Meso-Erythritol may be used in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications. It may be used as a reference compound in purification and analytical procedures developed to test fermentation broths.

Biochem/physiol Actions

Allelic variation of the Tas1r3 gene affects behavioral taste responses to this sugar alcohol, suggesting that it is a T1R3 receptor ligand.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karnjapan Janthawornpong et al.
Journal of the American Chemical Society, 135(5), 1816-1822 (2013-01-16)
The MEP pathway, which is absent in animals but present in most pathogenic bacteria, in the parasite responsible for malaria and in plant plastids, is a target for the development of antimicrobial drugs. IspH, an oxygen-sensitive [4Fe-4S] enzyme, catalyzes the
H Hino et al.
Acta dermato-venereologica, 62(3), 185-191 (1982-01-01)
Keratinocytes were dissociated from normal human adult epidermis with clostridial collagenase, dithio-erythritol and trypsin, and cultured. Immediately after this, no connection was seen between contiguous cells. Ruptured desmosomes, with masses of tonofilaments and distinct attachment plaques were still left on
Man-Yeon Choi et al.
Journal of economic entomology, 112(2), 981-985 (2018-11-30)
Previous studies have demonstrated various combinations of non-nutritive erythritol and sucrose having detrimental effects on Drosophila suzukii (Matsumura). Fly mortality is likely caused by 1) starvation from feeding on non-metabolizable erythritol; and 2) physiological imbalance with abnormally high osmotic pressure
K Noda et al.
European journal of clinical nutrition, 48(4), 286-292 (1994-04-01)
To investigate the effect of an oral administration of erythritol on serum glucose and insulin levels in healthy subjects and estimate available energy of erythritol in human. Ingestion of erythritol (0.3 g/kg body weight) or the same dose of glucose
Yanmei Xiao et al.
Cell, 149(7), 1525-1535 (2012-06-26)
Plastid-derived signals are known to coordinate expression of nuclear genes encoding plastid-localized proteins in a process termed retrograde signaling. To date, the identity of retrograde-signaling molecules has remained elusive. Here, we show that methylerythritol cyclodiphosphate (MEcPP), a precursor of isoprenoids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service