E7625
D-(−)-Erythrose
≥75% (TLC), syrup
Synonym(s):
(2R,3R)-2,3,4-trihydroxybutanal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
Beilstein:
1721698
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥75% (TLC)
form
syrup
color
colorless to very dark yellow
storage temp.
2-8°C
SMILES string
[H]C(=O)[C@H](O)[C@H](O)CO
InChI
1S/C4H8O4/c5-1-3(7)4(8)2-6/h1,3-4,6-8H,2H2/t3-,4+/m0/s1
InChI key
YTBSYETUWUMLBZ-IUYQGCFVSA-N
Looking for similar products? Visit Product Comparison Guide
Application
D-(-)-Erythrose, the D enantiomer of the aldose aldehyde erythrose, may be used as a reference compound in sugar metabolism analysis. D-(-)-Erythrose may be used to help identify and characterize erythrose reductase(s). D-Erythrose may be used to study the mechanisms of mutarotation in monosugars. D-Erythrose may be used to study the mechanisms of organic microspherule formation and Maillard (glycation) reactions.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Birgit Jovanović et al.
AMB Express, 3(1), 43-43 (2013-08-09)
Proteins with putative erythrose reductase activity have been identified in the filamentous fungi Trichoderma reesei, Aspergillus niger, and Fusarium graminearum by in silico analysis. The proteins found in T. reesei and A. niger had earlier been characterized as glycerol dehydrogenase
Tomasz Janek et al.
Microbial cell factories, 16(1), 118-118 (2017-07-12)
Erythritol is a natural sweetener that is used in the food industry. It is produced as an osmoprotectant by bacteria and yeast. Due to its chemical properties, it does not change the insulin level in the blood, and therefore it
Danielle Rand et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 41(1), 17-22 (2010-06-22)
Reactions of short sugars under mild, plausibly prebiotic conditions yield organic microspherules that may have played a role in prebiotic chemistry as primitive reaction vessels. It has been widely thought that nitrogen chemistry, in particular Amadori rearrangement, is central to
Dae-Hee Lee et al.
Microbial cell factories, 9, 43-43 (2010-06-10)
Erythrose reductase (ER) catalyzes the final step of erythritol production, which is reducing erythrose to erythritol using NAD(P)H as a cofactor. ER has gained interest because of its importance in the production of erythritol, which has extremely low digestibility and
Julia Schörghuber et al.
Journal of biomolecular NMR, 71(3), 129-140 (2018-05-29)
In recent years, we developed a toolbox of heavy isotope containing compounds, which serve as metabolic amino acid precursors in the E. coli-based overexpression of aromatic residue labeled proteins. Our labeling techniques show excellent results both in terms of selectivity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service