Skip to Content
Merck

Skip To

10065

α-Amylase from Aspergillus oryzae

powder, ~30 U/mg

Synonym(s):

1,4-α-D-Glucan-glucanohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-588-1
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
~30 U/mg
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

specific activity

~30 U/mg

mol wt

Mr ~51000

storage temp.

2-8°C

General description

Microbial amylases are exoenzymes and are used in several industrial applications, such as production of bread, maltose syrups, and fermentation of soya sauce, miso etc. α-Amylase isolated from Aspergillus oryzae has been found to have molecular weights of 51kDa (sedimentation and diffusion) and 49kDa (gel filtration).

Application

α-Amylase has been used:
  • as a control enzyme in agar plate-based and carboxymethylcellulose-based clearing assays to screen cellulase activity
  • for the hydrolysis of starch to explore the the role of wheat starch in frozen dough
  • to inhibit Staphylococcus aureus biofilms

Biochem/physiol Actions

Aspergillus oryzae α-amylase (Ao α-amylase) enzyme catalyzes the hydrolysis of the α-1,4 glycosidic bonds in soluble starches and related subsrates. These substrates are broken down to release short oligosaccharides and α-limit dextrins.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.0 and 25°C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate).
For the determination of fats in food; Application in (selective) hydrolysis/condensation of glycosidic bonds.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
86250A9857A8220
description

powder, ~30 U/mg

description

powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)

description

≥150 units/mg protein (biuret)

description

aqueous solution, ≥800 FAU/g

specific activity

~30 U/mg

specific activity

~1.5 U/mg (~0.2 U acc. to Willstätter)

specific activity

≥150 units/mg protein (biuret)

specific activity

≥800 FAU/g

form

powder

form

powder

form

powder

form

aqueous solution

mol wt

Mr ~51000

mol wt

-

mol wt

-

mol wt

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under α-Amylase from Aspergillus oryzae


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters