Skip to Content
Merck

A8220

α-Amylase from Aspergillus oryzae

greener alternative

aqueous solution, ≥800 FAU/g

Synonym(s):

Fungamyl® 800L, 1,4-α-D-Glucan-glucanohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-588-1
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
≥800 FAU/g
Biological source:
Aspergillus sp. (A. oryzae)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Aspergillus sp. (A. oryzae)

Quality Level

form

aqueous solution

specific activity

≥800 FAU/g

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

α-Amylase is also referred as taka-amylase A.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Application

α-Amylase from Aspergillus oryzae has been used:
  • as a positive control in the evaluation of enzyme activity
  • to evaluate its effect on simultaneous saccharification and cofermentation (SSCF) of L-lA by Lactobacillus paracasei LA104{65
  • to study the influence of amylase

  • α-Amylase are suitable various plant studies, such as metabolism studies in Arabidopsis .
  • It is a suitable positive control for the evaluation of exogenous enzymes with amylolytic activity for dairy cows.
  • Amylases from Aspergillus oryzae is a suitable baking additives that prevents staling in the baking industry, clarify haze from fruit juices and alcoholic beverages, and is used to produce glucose and maltose syrup products .

Biochem/physiol Actions

α-Amylase, a digestive enzyme in saliva helps to catalyse the hydrolysis of α-1,4 glycosidic linkages in starch and results in the formation of glucose units. It is effective at slightly alkaline pH and is one of the preferred enzymes for sake brewing.
Aspergillus oryzae α − amylase (Ao α-amylase) enzyme catalyzes the hydrolysis of the α-1,4 glycosidic bonds in soluble starches, and is used in various industrial applications. Natural substrates such as starch and glycogen are broken down into glucose and maltose.
.

Legal Information

A product of Novozyme Corp.
Fungamyl is a registered trademark of Novozymes Corp.


Still not finding the right product?

Explore all of our products under α-Amylase from Aspergillus oryzae


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



I Sander et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 37(8), 1229-1238 (2007-07-27)
In order to enable reproducible and comparable exposure measurements of fungal alpha-amylase (alpha-amylase) in different laboratories and countries, the entire procedure from sampling of airborne dust to measuring extracted samples (including standards and the used enzyme) immunoassays must be standardized.
Towards effective and stable probiotics
Yarullina DR, et al.
The International Journal of Risk & Safety in Medicine, S65-S66 (2015)
THE MOUTH, SALIVARY GLANDS AND OESOPHAGUS
Smith ME, et al.
Digestive System, 19-38 (2010)



Global Trade Item Number

SKUGTIN
A8220-50ML04061838591197
A8220-250ML04061833391334