Skip to Content
Merck
All Photos(3)

Key Documents

D137510

Sigma-Aldrich

N,N-Dimethylacetamide

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

vapor density

3 (vs air)

Quality Level

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide (DMAc) is a water-miscible organic solvent used in the synthesis of styrene′s, vinyl resins, linear polyesters, acrylic fibers, plastics, pesticides, and synthetic leather. Additionally, it is utilized in the production of adhesives, films, and paint. It participates in dehydrogenation, dehydration, halogenation, deprotonation reactions and reductive carbonylation of transition metals. It can also act as catalysts, promoters, halogen/alkoxy or hydroxyl/halogen exchange mediators, reducing agents, and sources of the base.

Application

N,N-Dimethylacetamide can be used as a solvent in the synthesis of:
  • Cellulose-HA (hydroxyapatite) nanocomposites using microcrystalline cellulose, CaCl2, and NaH2PO4.
  • 5-hydroxymethylfurfural (HMF) from various carbohydrates and natural biopolymer lignocellulose in the presence of LiCl. 

It can also be used:
  • In the Pd-catalyzed regioselective synthesis of ketones from olefins using molecular oxygen.
  • As a solvent as well as a methylene source in the regioselective linkage of two imidazo[1,2-a] pyridines using Cu(OAc)2 catalyst.

Features and Benefits

N,N-Dimethylacetamide has
  • High dissolution power of various substrate.
  • High capacity to solvate anions.

Legal Information

Belle Chemical Company
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
S Sood et al.
Animal reproduction science, 129(1-2), 89-95 (2011-11-15)
Three experiments conducted to improve the survival of emu sperm during cryopreservation aimed to: (1) minimize chilling injury during the cooling phase; (2) determine the osmotic effects of dimethylacetamide (DMA), sucrose and trehalose; and (3) investigate the timing and nature
Agnieszka Partyka et al.
Theriogenology, 75(9), 1623-1629 (2011-03-15)
The aim of this study was to perform flow cytometric analysis of C11-BODIPY581/591 oxidation in fowl and geese sperm as a marker for membrane lipid peroxidation (LPO) and to establish if the cryopreservation process would make sperm membranes more susceptible
Jingshuai Yang et al.
ChemSusChem, 6(2), 275-282 (2013-01-11)
Covalently cross-linked polymer membranes were fabricated from poly(aryl sulfone benzimidazole) (SO(2)PBI) and poly(vinylbenzyl chloride) (PVBCl) as electrolytes for high-temperature proton-exchange-membrane fuel cells. The cross-linking imparted organo insolubility and chemical stability against radical attack to the otherwise flexible SO(2)PBI membranes. Steady
Wesley Walker et al.
Journal of the American Chemical Society, 135(6), 2076-2079 (2013-01-31)
A major challenge in the development of rechargeable Li-O(2) batteries is the identification of electrolyte materials that are stable in the operating environment of the O(2) electrode. Straight-chain alkyl amides are one of the few classes of polar, aprotic solvents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service