Skip to Content
Merck
All Photos(3)

Key Documents

271012

Sigma-Aldrich

N,N-Dimethylacetamide

anhydrous, 99.8%

Synonym(s):

DMAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

grade

anhydrous

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

99.8%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

impurities

<0.005% water

evapn. residue

<0.001%

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylacetamide may be used as a solvent in the following processes:
  • Palladium (II) acetate-butyldi-1-adamantylphosphine catalyzed arylation of 2-isobutylthiazole with chlorobenzene to form 5-phenyl-2-isobutylthiazole.
  • Synthesis of β-, γ-, and δ-metallo ester intermediates that can react with acid chlorides to form γ-, δ-, and ε-keto esters.
  • Synthesis of (R)-(-)-methyl 1,1′-binaphthyl-2,2′-diylphosphate that can be used as a precursor for preparing (R)-(+)-1,1′-binaphthalene-2,2′-diol.

Other Notes

The article number 271012-4X2L will be discontinued. Please order the single bottle 271012-2L which is physically identical with the same exact specifications.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Density and Viscosity Measurements for Binary Mixtures of 1-Ethyl-3-methylimidazolium Tetrafluoroborate ([Emim][BF4]) with Dimethylacetamide, Dimethylformamide, and Dimethyl Sulfoxide.
Fan XH, et al.
Journal of Chemical and Engineering Data (2016)
(R)-(+)-1,1'-Binaphthalene-2,2'-diol
Truesdale LK
Organic Syntheses, 67, 13-13 (1989)
Ethyl 5-oxo-6-methyl-6-heptenoate from methacryloyl chloride and ethyl 4-iodobutyrate
Yoshinao T, et al
Organic Syntheses, 67, 98-98 (1989)
Solutions of cellulose in N, N-dimethylacetamide/lithium chloride studied by light scattering methods.
Roder T, et al.
Polymer, 42(16), 6765-6773 (2001)
Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service