Skip to Content
Merck
All Photos(3)

Key Documents

B9876

Sigma-Aldrich

Sodium tetraborate decahydrate

Sodium tetraborate decahydrate

ReagentPlus®, ≥99.5%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder
Pricing and availability is not currently available.

grade

reagent

Agency

suitable for SM 4500 - NH3

product line

ReagentPlus®

Assay

≥99.5%

form

powder

pH

9.2 (10 g/L)

density

1.73 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium tetraborate decahydrate is widely used to prepare buffer solution with a pH range approx 9.3. It can also be used as:
  • A component of a buffer solution used in the separation and estimation of thiamin and its phosphate esters in commercial tablets.[1]
  • A fluxing agent in the preparation of ceramizable silicone rubber-derived composite materials.[2]
  • A crosslinking agent in the synthesis of porous foams by reacting cellulose nanofibres with poly-vinyl alcohol.[3]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of thiamin and its phosphate esters by capillary zone electrophoresis and its application to the analysis of water-soluble vitamins
Shabangi M and Sutton JA
Journal of Pharmaceutical and Biomedical Analysis, 38, 66-71 (2005)
Structural packaging foams prepared by uni-directional freezing of paper sludge cellulose nanofibres and poly (vinyl alcohol)
Adu C, et al.
Materials Letters, 253, 242-245 (2019)
Improving the mechanical and electrical properties of ceramizable silicone rubber/halloysite composites and their ceramic residues by incorporation of different borates
Guo J, et al.
Polymers, 10, 388-388 (2018)
C Cencetti et al.
Carbohydrate polymers, 90(3), 1362-1370 (2012-09-04)
Silver-loaded dressings are designed to provide the same antimicrobial activity of topical silver, with the advantages of a sustained silver release and a reduced number of dressing changes. Moreover, such type of dressing must provide a moist environment, avoiding fiber
Qingzhou Cui et al.
The Analyst, 134(5), 875-880 (2009-04-22)
We developed a photonic crystal sensing method for diol containing species such as carbohydrates based on a poly(vinyl alcohol) (PVA) hydrogel containing an embedded crystalline colloidal array (CCA). The polymerized CCA (PCCA) diffracts visible light. We show that in the

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service