Skip to Content
Merck

Skip To

B6768

Boric acid

BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
PubChem Substance ID:
eCl@ss:
38120104
UNSPSC Code:
12161700
NACRES:
NA.25
EC Number:
233-139-2
MDL number:
Assay:
≥99.5%
Concentration:
≥99.5%
Form:
powder
Grade:
Molecular Biology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

vapor pressure

2.6 mmHg ( 20 °C)

product line

BioReagent

assay

≥99.5%

form

powder

concentration

≥99.5%

technique(s)

cell culture | mammalian: suitable, cell culture | plant: suitable

pH

5.1 (25 °C, 1.8 g/L)

mp

160 °C (dec.) (lit.)

solubility

water: 40 mg/mL, clear, colorless

density

1.49 g/cm3 at 23 °C

cation traces

Fe: ≤5 ppm, Mg: ≤5 ppm, heavy metals (as Pb): ≤10 ppm

UV absorption

λ: 260 nm Amax: 0.05, λ: 280 nm Amax: 0.05

foreign activity

DNase, RNase, and protease, none detected

storage temp.

room temp

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

General description

Boric acid is a weak monobasic Lewis acid (electron pair acceptor), which reacts reversibly with alcohols. It is used in dermal and women′s hygiene products due to its mild antibacterial and antifungal effects. Boric acid is used as a buffer in ophthalmic products. It acts as an effective solid lubrication agent because of its molecular structure. Boric acid finds its applications in glass making, fire retardant. Boric acid is used in biochemistry in buffer solutions. It also forms complex equilibria in solution, with the tetraborate ion (B4O7 2- ) being the predominant species. Borate buffers are involved in discontinuous electrophoresis and capillary electrophoresis systems for the separation of proteins and nucleic acids.

Application

Boric acid has been used:
  • to prepare boric buffer for the quantification of primary amine groups at the surface of nanocapsules
  • to study its effects on epigenetic change
  • for the estimation of protein in wheatgrass using Kjeldahl method

Used to promote amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.
Intensified Azeotropic Distillation: A Strategy for Optimizing Direct Amidation

Features and Benefits

  • Environment-friendly
  • Non-toxic 


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
31146B7901202878
grade

Molecular Biology

grade

ACS reagent, puriss. p.a.

grade

electrophoresis grade

grade

-

concentration

≥99.5%

concentration

-

concentration

-

concentration

-

assay

≥99.5%

assay

≥99.8%

assay

≥99.5%

assay

99.999% trace metals basis

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder or crystals

form

powder

form

crystalline powder

pH

5.1 (25 °C, 1.8 g/L)

pH

3.6-4.0 (20 °C, 4%)

pH

8.2-10.1

pH

-


Still not finding the right product?

Explore all of our products under Boric acid


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. Hello, I am working on a tissue clearance project with this product. From the literature review, I noticed the use of 4% (wt/vol) SDS to 400mM borate as a crucial step. In the literature, there are ambiguous references to either borate or boric acid.

    1 answer
    1. The word borate is probably being used to describe a borate buffer. Borate buffers are typically used around pH 9. One could prepare a sodium borate buffer by dissolving this boric acid product in water (to give a solution at approximately pH 5) followed by the addition of sodium hydroxide to bring it up to pH 9. But the more typical approach is to use Sodium tetraborate decahydrate (e.g., Product B9876). Dissolving this Sodium tetraborate decahydrate in water will give a solution that has a pH of approximately 9.3. Then, the Sodium dodecyl sulfate (SDS) could be added to that solution.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. How can I find out the expiration date? Lot No : BCCH1940

    1 answer
    1. Expiration information will be on the certificate of analysis. The certificate of analysis can be accessed by clicking on the "COO/COA" tab under the product photo the product detail page or the scrolling down to the documentation section. Here is a copy of lot BCCH1940 COA: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/B6768-BULKBCCH1940.pdf.

      This product does not contain an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services

      Helpful?

Reviews

No rating value

Active Filters