Skip to Content
Merck
All Photos(1)

Key Documents

95662

Sigma-Aldrich

o-Xylene

puriss. p.a., ≥99.0% (GC)

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (GC)
bp:
143-145 °C (lit.)
vapor pressure:
<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

vapor density

3.7 (vs air)

Quality Level

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

867 °F

expl. lim.

7 %

impurities

≤0.4% toluene (GC)
≤0.6% m- and p-xylene (GC)

evapn. residue

≤0.002%

refractive index

n20/D 1.505 (lit.)
n20/D 1.505

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

solubility

water: partially soluble 0.1705 g/L at 25 °C

density

0.879 g/mL at 20 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Xylene is an isomeric form of xylene. Experimental values of density (ρ) and speed of sound (u) of its binary mixtures with diethyl carbonate (DEC) have been evaluated over the temperature range of 298.15-323.15K and at 0.1MPa atmospheric pressure. Its oxidation in the presence of three-dimensionally ordered mesoporous Co3O4 (meso-Co3O4) supported gold (xAu/meso-Co3O4, x = 3.7-9.0wt%) nanocatalysts has been reported.

Application

o-Xylene may be used in the synthesis of phthalic anhydride and phthalodinitrile.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous Co3O4-supported gold nanocatalysts: Highly active for the oxidation of carbon monoxide, benzene, toluene, and o-xylene.
Liu Y, et al.
J. Catal., 309, 408-418 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 833-833 (1994)
Casper Bindzus Foldager et al.
Cartilage, 5(2), 123-132 (2014-04-01)
The objective of the present study was to investigate the presence and distribution of 2 basement membrane (BM) molecules, laminin and collagen type IV, in healthy and degenerative cartilage tissues. Normal and degenerated tissues were obtained from goats and humans
Influence of alkyl group and temperature on excess thermodynamic properties of diethyl carbonate and their binary mixtures at 0.1 MPa.
Srilakshmi M, et al.
Journal of Molecular Liquids, 211, 854-867 (2015)
Maia Chanrion et al.
Nature communications, 5, 5005-5005 (2014-10-09)
Epithelial-to-mesenchymal transition-like (EMT-like) is a critical process allowing initiation of metastases during tumour progression. Here, to investigate its role in intestinal cancer, we combine computational network-based and experimental approaches to create a mouse model with high metastatic potential. Construction and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service