Skip to Content
Merck
All Photos(1)

Key Documents

295884

Supelco

o-Xylene

suitable for HPLC, 98%

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
143-145 °C (lit.)
vapor pressure:
<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

grade

HPLC grade

vapor density

3.7 (vs air)

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

867 °F

purified by

glass distillation

expl. lim.

7 %

technique(s)

HPLC: suitable

impurities

<0.020% water

evapn. residue

<0.0003%

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

solubility

water: soluble 0.1705 g/L at 25 °C

density

0.879 g/mL at 20 °C (lit.)

λ

H2O reference

UV absorption

λ: 288 nm Amax: 1.00
λ: 300 nm Amax: 0.40
λ: 325 nm Amax: 0.05
λ: 350-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Photodegradation of xylene isomers: The study explores the kinetics, mechanism, secondary pollutant formation potential, and health risk evaluation associated with the photodegradation of xylene isomers, providing insights into environmental and safety aspects relevant to its use (Chen et al., 2024).

  • Green-Solvent Processed Blade-Coating Organic Solar Cells: Demonstrates the use of green solvents in the fabrication of organic solar cells, achieving an efficiency close to 19%, which underscores o-Xylene′s relevance in developing environmentally friendly solvent systems for high-performance solar cells (Bai et al., 2023).

  • Application of Composts′ Biochar as Potential Sorbent: Highlights the application of biochar derived from composts in reducing VOCs emission during kitchen waste storage, suggesting a viable application of o-Xylene in environmental management strategies (Rosik et al., 2023).

  • A-A Strategy Enables Desirable Performance of All-Polymer Solar Cells: Discusses a novel approach to fabricating all-polymer solar cells using nonhalogenated solvents, promoting the use of o-Xylene in creating more sustainable and efficient photovoltaic devices (Peng et al., 2023).

  • Understanding the Thickness and Light-Intensity Dependent Performance: This study addresses the performance variables in green-solvent processed organic solar cells, contributing to the optimization of solar energy technologies where o-Xylene′s solvent properties may be utilized (Lübke et al., 2023).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous Co3O4-supported gold nanocatalysts: Highly active for the oxidation of carbon monoxide, benzene, toluene, and o-xylene.
Liu Y, et al.
J. Catal., 309, 408-418 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 833-833 (1994)
Casper Bindzus Foldager et al.
Cartilage, 5(2), 123-132 (2014-04-01)
The objective of the present study was to investigate the presence and distribution of 2 basement membrane (BM) molecules, laminin and collagen type IV, in healthy and degenerative cartilage tissues. Normal and degenerated tissues were obtained from goats and humans
Pawaree Saisawat et al.
Kidney international, 85(6), 1310-1317 (2013-10-25)
Congenital abnormalities of the kidney and urinary tract (CAKUT) account for approximately half of children with chronic kidney disease and they are the most frequent cause of end-stage renal disease in children in the US. However, its genetic etiology remains
Influence of alkyl group and temperature on excess thermodynamic properties of diethyl carbonate and their binary mixtures at 0.1 MPa.
Srilakshmi M, et al.
Journal of Molecular Liquids, 211, 854-867 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service